log in or register
Insulated Static Discharge Stick, 4 foot length, with 6 foot grounding cable and clamp with strain relief
EW-86437-01
$477.00 USD/EACH
Usually ships in 10 days.
Qty
  • Remove static charge after de-energizing a circuit to improve worker safety
Safely remove static charge after de-energizing with these static discharge sticks. Stick comes preassembled with a brass alloy U-hook and is constructed with closed cell foam-filled tubular fiberglass, made in accordance with ASTM F711. Includes 6-ft (1.8 m) of copper grounding cable attached to a flat jaw grade 3 bronze serrated clamp with strain relief.
Model 20817
Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title