Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

Isopropyl Alcohol Acs 1L

EW-89000-91
$29.00 USD/EACH

quantity discounts Quantity Discounts
Order Discount Price
[X]
6 10% $26.10/EACH
12 15% $24.65/EACH
24 20% $23.20/EACH
Usually ships in 5 days.
Qty

This item is considered a hazardous material.Please note: there may be additional shipping restrictions like freight collect or no priority air service, extra freight charges may apply. This item(s) may be restricted from shipping to a residential address, sold to private individuals or exported from the United States. For additional information or questions please call us at 1-800-323-4340 (US) or 847-549-7600 (International).
Product Type Chemical Reagents
Purity grade ACS
CAS Number 67-63-0
Form Liquid
Formula weight 60.10
Empirical formula C3H8O
Linear formula (CH3)2CHOH
Synonyms 2-Propanol, IPA, Isopropanol
Packaging Amber Glass Bottle
Storage symbol Red
UN number UN 1219
Hazard symbol Flammable
Quantity (mL) 1000
DOT information DOT 3 PG II
DOT description Isopropanol
Brand Cole-Parmer
Manufacturer number 900091
Quantity 1 L





Technical Resources
Certificate of Analysis
Enter lot number

or click to view a Sample C of A
 
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title