Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Jonard GD-50 Precision Dynamometer; 60-500 g
JONARD INDUSTRIES CORP  |   Mfr# GD-50  |   Item# EW-93955-14
$370.00 USD/EACH
Are you a reseller or buying in bulk? Call us today.
Usually ships in 7 days.
Qty
    Measurement is extremely simple. Just place point of gauge arm perpendicular to the force to be measured. Reading can be taken in both directions- clockwise and counter-clockwise.
    All Gauges Equipped With Maximum Reading Pointer
    Maximum reading pointer moves with indicator when testing. It will remain at maximum force measured, enabling user to take reading afterward. No need to look at dial hand while measuring, thus avoiding reading errors.
    Gauges Offer Extreme Precision And Durability
    The measuring springs are made of hard-tempered annealed beryllium copper. This alloy provides outstanding flexibility with minimum effort. Dial is protected by unbreakable glass. Packaged in rigid, fitted plastic case to protect instrument.
    All gauges equipped with maximum reading pointer…
    (Reading + Maximum Scale Reading)
    APPLICATIONS
    • Spring tension of all relay contacts and switches
    • Starting torque or motors, clock movements, etc.
    • Pressure of brushes in electro-mechanical devices, phono tone arms, needles, etc.
    Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
    Product Type Dynamometers
    Description Precision Dynamometers
    Model GD-50
    Qty/ea 1
    Brand JONARD INDUSTRIES CORP
    Display Analog dial with rotational tare ring
    Manufacturer number GD-50
    Calibrations
    Qty/Price
    Availability
    at $217.00 USD/EACH
    Usually ships in 11 days.
    Technical Resources
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456