United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 4
of
6 >
EW-81791-44
81791-45 shown
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-81791-44
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Safety Products
>
Storage and Handling
>
Safety Cans for Flammables
Share
|
Justrite Safety Can for Flammables, Type II, 1 gallon
EW-81791-44
$90.00 USD/EACH
Call 1-888-358-4705 for availability.
Qty
:
Add to Cart
Save to shopping list
Safely contain hazardous liquids and control vapors
A necessary part of compliance for safety in the workplace
Reduce the risk of fire, protecting both personnel and facilities
Improve efficiency with ease-of-use features to enhance pouring and filling operations
All cans include a stainless steel flame arrester that prevents flashback ignition to reduce fire risks; also include a large ID zone for user labeling of contents to avoid misuse. Self-closing caps are designed with a relief mechanism to vent pressureprotects against rupture and explosion.
Type II Safety Cans
are perfect for repetitive use applications to reduce worker strain and fatigue. The "one-spout" design features a Safe-Squeeze
®
trigger mechanism and AccuFlow
manifold, offering a highly controlled liquid flow method. As the trigger is engaged, the piston-actuated spout and vacuum release valve open simultaneously for accurate, glug-free pouring. Flexible 9"L metal hose ensures easy positioning for smaller openings.
Approval: FM, UL, cUL
Compliance: OSHA, NFPA
Specifications
Product Type
Safety Cans
Capacity (Liters)
4
Capacity (gal.)
1
Dimensions (in.)
10.5 in H x 9.5 in dia. (26.7 x 24.1 cm)
Material
Steel
Brand
Justrite
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title