United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
EW-10382-16
Kestrel 3000 Pocket Wind Meter; Measures Wind Speed Temperature Windchill Humidity Heat Stress And Dew Point
Click to enlarge
More Product Details
Specifications
Accessories
Calibrations
Technical Resources
Instruction manual
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-10382-16
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17101-01
NIST-Traceable Calibration; Anemometers, Vane-Type
$225.00 USD/EACH/EACH
Usually ships in 13 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Temperature, Humidity, Moisture and Weather Products
>
Weather Products
>
Handheld Weather Instruments
>
Handheld Weather Instruments Conditions Monitoring
>
Kestrel 3000 Pocket Wind Meter
Share
|
Kestrel 3000 Pocket Wind Meter; Measures Wind Speed Temperature Windchill Humidity Heat Stress And Dew Point
EW-10382-16
$199.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Measure air velocity and other weather parameters accurately and easily
Wide range of field selectable units
Large easy-to-read display
Waterproof IP67-rated casing
These convenient handheld meters will measure wind speed and other weather parameters with field selectable units including:
Knots (KT)
Meters per second (M/S)
Kilometers per hour (KM/H)
Miles per hour (MPH)
Feet per minute (FPM)
Beaufort force("B")
KESTREL 3000 - Pocket Weather Meter Features
Wind Speed
Temperature
Wind Chill
Humidity
Heat Stress
Dew Point
A number of accessories are available including a portable vane mount and a tripod for longer readings, and a replacement impeller.
What's Included:
Slip-on protective cover, neck lanyard, and CR2032 battery.
Specifications
Product Type
Vane Anemometer
Humidity range
0.0 to 100.0%, 5.0 to 95.0% noncondensing
Dew point range
15 to 95%
Temp accuracy
0.9°F (0.5°C)
Temp range
-20.0 to 158.0°F (-29.0 to 70.0°C)
RH range
5 to 95%
RH accuracy
0.03
Dew point accuracy
3.4°F (1.9°C)
Wind speed range
118 to 7874 ft/min
Display
3-1/2 digit LCD, aviation green backlight
Dimensions
4-7/8" L x 2" W x 1-1/8" H (12.2 x 4.8 x 2.8 cm)
Power
CR2032 battery
CE Compliance
Yes
Brand
Kestrel
Manufacturer number
0830
Accessories
Qty
/
Price
Availability
Kestrel 0802 RH Calibration Kit
EW-10382-12
at
$99.00 USD/EACH
Usually ships in 7 days.
Add to Cart
Kestrel 0801 Replacement Impeller for Kestrel Pocket Instruments
EW-10382-13
at
$19.00 USD/EACH
Usually ships in 6 days.
Add to Cart
Kestrel 0791 Portable Vane Mount for Kestrel Pocket Instruments
EW-99152-61
at
$69.00 USD/EACH
Usually ships in 5 days.
Add to Cart
Kestrel 0807 Portable Tripod for Kestrel Pocket Instruments
EW-99152-62
at
$25.00 USD/EACH
Usually ships in 5 days.
Add to Cart
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; Anemometers, Vane-Type
EW-17101-01
at $225.00 USD/EACH
Usually ships in 13 days.
Add to Cart
Technical Resources
kestrel meter specificationss.pdf
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Temperature FAQs
Temperature Instrument Ranges
Getting a Handle on Temperature
Show All...
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title