United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 4
of
4
Representative photo only
Kimcote Heavy Wall Filter Flask 4000mL
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-34524-55
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Laboratory Supplies & Consumables
>
Flasks
>
Filtering Flasks
>
Glass Filtering Flasks
>
Kimcote, Heavy-Wall, Glass Safety Coated, Filtering Flasks
Share
|
Kimcote, Heavy-Wall, Filter Flask, 4000mL
EW-34524-55
$209.00 USD/CASE
In stock.
Qty
:
Add to Cart
Save to shopping list
Heavy wall, filtering
Ultra-clear safety-coated glassware helps stop and control spills in the event of glassware breakage. If a break does occur, the durable odor-free glassware will reduce the hazards of shattered glass and leakage of toxic or corrosive chemicals. Glassware is ideal for chemical storage, transportation, and handling of hazardous chemicals, harmful biologicals, and acids. Glassware has a unique nonslip surface when wet or dry. Heavy wall borosilicate glass offers mechanical strength for vacuum filtration. Flask has a #2 side arm for connection to ¼” (6mm) ID flexible tubing. Autoclavable to120°C (250°F).
Specifications
Capacity (mL)
4000
Material
Glass
Stopper size
8
Height (cm)
38.0
Quantity
1
Brand
Kimcote
Manufacturer number
953760-4002
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title