dummy
EW-98928-25
Call 1-888-358-4705 for availability.
Qty:
- Microprocessor based bath provides high-accuracy temperature control
- Conforms to ASTM D 88 and ASTM E 102
Constant temperature bath meets ASTM requirements for precise temperature control. The microprocessor PID control offers quick stabilization without overshooting; overtemperature cutoff further protects bath and application from exceeding the cutoff point.
The heavy-gauge stainless steel bath interior is insulated. The chemically-resistant top plate provides further insulation and is removable for easy cleaning of bath interior.
Digital display provides actual and set temperature indication in both °C and °F. Recommended bath fluid is water or silicone heat transfer fluid. For near ambient temperatures, circulate tap water or coolant through the built-in cooling coil. For your specific test method needs, order an ASTM glass thermometer separately from our temperature section.
| Product Type |
Viscosity Bath |
| Bath type |
Viscosity Water Bath |
| Capacity (Liters) |
7 |
| Min temperature (º C) |
Ambient |
| Max temperature (º C) |
240 |
| Power (VAC) |
230 |
at $352.00 USD/EACH
Call 1-888-358-4705 for availability.
at $867.00 USD/EACH
Call 1-888-358-4705 for availability.
at $70.00 USD/EACH
Call 1-888-358-4705 for availability.
at $145.00 USD/EACH
Usually ships in 9 days.
at $78.00 USD/EACH
Usually ships in 15 days.
Technical Resources
Related technical articles (Roll over for summary)
Show All...
Show Less...
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334