log in or register
LaMotte 2020we Handheld Turbidity Meter, EPA compliant
EW-05563-22
$979.00 USD/EACH
  • Redesigned for higher accuracy and wider range
  • Advanced microprocessor offers high-accuracy readings
  • Available with either EPA- or ISO-compliant optical systems
  • Battery operated for field and on-site testing convenience
Test municipal waters, food and beverage waters, or any aqueous solution where fluid clarity is important. These compact portable meters are equipped with a unique optics configuration to read low turbidity levels down to 0.05 NTU and high levels up to 4000 NTU. Large, menu-driven LCD features five measuring units, seven user-selectable languages, datalogging capabilities to 500 points, and USB interface for downloading data to your computer.
What's Included:
60 mL each of 0, 1, and 10 NTU standards; plastic sample bottle; four glass sample vials; USB cable and wall plug; and waterproof plastic carrying case.
Product Type Turbidity Meter
Meter style Portable
Dimensions (" W) 3-1/2
Dimensions (" H) 2-1/2
Dimensions (" L) 7-1/2
Range (Turbidity) 0 to 4000
Range (Temperature °C) 0 to 50
Range (Temperature °F) 32 to 122
Resolution (Turbidity) 0.01, 0.1, 1
Accuracy (Turbidity) ± 2% of reading
Display LCD
Interface USB
Waterproof Yes
Power Rechargeable lithium battery (included)
Datalogging (points) 500
Brand LaMotte
Model 1970-EPA
Accessories
Qty/Price
Availability
at $47.00 USD/EACH
In stock.
at $47.00 USD/EACH
In stock.
at $47.00 USD/EACH
In stock.
at $29.00 USD/PACKAGE
In stock.
at $29.25 USD/EACH
In stock.
Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    Customers Who Bought This Item Also Bought
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title