log in or register
LaMotte LTC3000we Benchtop Turbidity and Chlorine Meter, EPA compliant
EW-05563-30
$1,350.00 USD/EACH
  • Measure turbidity and chlorine with one convenient instrument—no need for multiple meters
  • Optical system permits EPA-compliant measurements for turbidity and chlorine
  • Innovative measurement technology provides added precision, sensitivity, and dependability
  • Simple user interface and large LCD make the meter easy to use
This innovative water analysis instrument is designed for precision measurement of turbidity and chlorine, making it fit well into a variety of lab applications. Use meter for testing drinking water, food and beverage waters, or for any other solutions where sample clarity is important. Advanced optics allow for measurements that are compliant with U.S. EPA 180.1 for turbidity and EPA 330.5 for chlorine.
Meter features intuitive menu-driven operation via an easy-to-read LCD. Six selectable languages, datalogging capabilities, and programmable auto-off provide added convenience
What's Included:
60 mL each of 0, 1, 10 NTU standards; 100 each of free and total chlorine DPD tablets; sample bottle; four sample tubes; and universal AC adapter.
Product Type Turbidity Meter
Meter style Benchtop
Dimensions (" W) 8-3/4
Dimensions (" H) 3
Dimensions (" D) 7-3/4
Range (Turbidity) 0 to 4000
Range (ISE) 0 to 10 ppm (chlorine)
Resolution (Turbidity) 0.01, 0.1, 1
Resolution (ISE) 0.01 ppm from 0 to 5 (chlorine)
0.1 ppm from 5 to 10 (chlorine)
Accuracy (Turbidity) ± 2% of reading
Accuracy (ISE) 0.05 or ±2% (chlorine)
Display LCD
Interface RS-232
Waterproof Yes
Power 110/220 VAC
Datalogging (points) 4,000
Brand LaMotte
Model 1965-EPA
Accessories
Qty/Price
Availability
at $50.00 USD/EACH
Usually ships in 8 days.
at $12.50 USD/PACKAGE
Usually ships in 8 days.
at $29.00 USD/PACKAGE
In stock.
at $13.00 USD/BOX
In stock.
at $13.25 USD/BOX
In stock.
at $29.25 USD/EACH
In stock.
at $49.00 USD/EACH
Call 1-888-358-4705 for availability.
at $49.00 USD/EACH
Usually ships in 9 days.
at $50.00 USD/EACH
Usually ships in 9 days.
Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title