United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 2
of
4 >
Representative photo only
LaMotte Single Test Colorimeter Kit Chlorine free and total in liquid form
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Instruction manual
Virtual Catalog (PDFs)
MSDS
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-99562-12
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
pH Meters and Water Quality Products
>
Colorimetry
>
Colorimeters
>
LaMotte Single Test Colorimeter Kits
Share
|
LaMotte Single Test Colorimeter Kit, Chlorine free and total in liquid form
EW-99562-12
$369.00 USD/EACH
Usually ships in 5 days.
Qty
:
Add to Cart
Save to shopping list
Single test, direct-reading colorimeters feature microprocessor technology
Analyzer features automatic wavelength selection for quick testing
Auto-zeroing and a hinged light cover make these meters easy to use. Ergonomic design fits in your hand comfortably. Water-resistant design protects your meter from damage. Download data to a computer or printer via the optional RS-232 interface.
What's included:
analyzer, glass sample vials, plastic water bottle, tablet crusher, test-specific reagent, foam-lined carrying case, and one 9 V battery.
This item is considered a hazardous material.
Please note: there may be additional shipping restrictions like freight collect or no priority air service, extra freight charges may apply. This item(s) may be restricted from shipping to a residential address, sold to private individuals or exported from the United States. For additional information or questions please call us at 1-800-323-4340 (US) or 847-549-7600 (International).
Specifications
Product Type
Color/Gloss Meters
Test parameters
Single parameter
Display
LCD
CE Compliance
Yes
Light source
LED
Detector
Silicon Photodiode
Datalogging
Yes
Dimensions
Meter: 6.5" L x 3.25" W x 2.5" H
Carrying case: 13.5" L x 9" W x 4.5" H
Tests/kit
144
Model
3670-01-LI
Brand
LaMotte
Manufacturer number
3670-01-LI
Power
one 9 V battery
Accessories
Qty
/
Price
Availability
RS-232 interface cable; RS-232 to 9-pin DIN connector, 10-ft L
EW-99562-56
at
$60.00 USD/EACH
Call 1-888-358-4705 for availability.
Add to Cart
Replacement test kit, chlorine free/total, liquid.
EW-99562-72
at
$18.00 USD/EACH
In stock.
Add to Cart
Technical Resources
Instruction Manual (616 KB)
MSDS 1 (135 B)
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Testing the pH of Wastewater Samples
Testing the pH of Soil Samples
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title