log in or register
Lab Companion 25L Reciprocal Shaking Water Bath - 120VAC, 60Hz
EW-12054-10
$3,320.00 USD/EACH
Usually ships in 6 days.
Qty
  • PID temperature and speed control at economical prices
  • Gentle start shaking minimzes shock to samples
  • Dual LED displays show actual or set temperature and shaking speed
  • Included gable cover minimizes fluid loss from evaporation
These shaking baths feature microprocessor temperature control with built-in calibration. Store up to three temperature settings for fast recall. Shaker mechanism has its own control key enabling you to use the unit as a constant temperature bath. Digital timer can be set from 1 to 99 hr 59 min and includes a delayed on/off capability. Shaking carriage can be removed allowing you to attach racks without contacting the fluid. Safety features include independent overtemperature and low-level alarms.
What's Included:
Stainless steel gable cover, spring wire rack, and 6-ft power cord with plug.
Product Type Water Bath
Bath type Shaking Water Bath
Capacity (Liters) 25
Min temperature (º C) Ambient
Max temperature (º C) 100
Temp control PID
Temp stability ±0.1°C at 50°C
Temp setting Splash-proof keypad
Low-level cutoff Yes
Power (VAC) 120
Power (Hz) 60
Timer 1 min to 99 hr 59 min
Accessories
Qty/Price
Availability
at $265.00 USD/EACH
Usually ships in 10 days.
at $17.00 USD/EACH
Usually ships in 10 days.
at $19.00 USD/EACH
Usually ships in 7 days.
at $26.00 USD/EACH
In stock.
at $34.00 USD/EACH
Usually ships in 10 days.
at $42.00 USD/EACH
Usually ships in 10 days.
at $67.00 USD/EACH
Usually ships in 5 days.
at $322.00 USD/EACH
Call 1-888-358-4705 for availability.
at $322.00 USD/EACH
Call 1-888-358-4705 for availability.
at $322.00 USD/EACH
Call 1-888-358-4705 for availability.
at $325.00 USD/EACH
Call 1-888-358-4705 for availability.
at $325.00 USD/EACH
Call 1-888-358-4705 for availability.
Calibrations
Qty/Price
Availability
at $250.00 USD/EACH
Usually ships in 21 days.
Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Leasing Available
    Financing is avaliable for many of our products.
    See Details
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title