United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths & Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cleaning & Janitorial Supplies
Cooling Equipment
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Lab Supplies & Consumables
Laboratory Equipment
Life Science
Masterflex® Peristaltic Pumps
Masterflex® Pump Tubing
Material Storage & Handling
Material Testing & Physical Testing
Microscopes & Magnification
Mixers and Blenders
pH, Conductivity, Dissolved Oxygen, and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity and Moisture Products
Tools
Tubing, Fittings, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
7 >
EW-32986-50
Lab-Scale Ultrasonic Flowmeter/Transmitter; pulse output, 0.001 to 0.01 GPM
Representative photo only
Click to enlarge
More Product Details
Specifications
Calibrations
Accessories
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-32986-50
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17080-12
NIST-Traceable Calibration; Liquid Flowmeter, 1 ccm to 300 LPM water
$250.00 USD/EACH/EACH
Usually ships in 15 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Flow, Level & Valves
>
Flowmeters
>
Ultrasonic Flowmeters
>
Lab-Scale Ultrasonic Flowmeters/Transmitters
Share
|
Sorry, this item is now discontinued.
Please browse similar items.
or
Contact us for more information.
Lab-Scale Ultrasonic Flowmeters/Transmitters
-
Lab-Scale Ultrasonic Flowmeter/Transmitter; pulse output, 0.001 to 0.01 GPM
EW-32986-50
$2,462.00 USD/EACH
Discontinued
Qty
:
Save to shopping list
Ideal for measuring high-purity solutions, slurries and corrosive chemicals
Flow through design with no intrusions produces almost no pressure loss
No mechanical seals to leak or moving parts to wear out
Segregated design for remote placement of sensor or transmitter/display
This design is ideal for use with fluids that have physical or chemical properties different from general-use water. The sensor utilizes PFA (perfluoroalkoxy) tubing, which is a high-purity, chemically inert fluoropolymer much like PTFE or FEP. The sensor housing is made of durable ABS plastic for placement in aggressive environments. A 6-ft (1.85m) cable allows for distant installation of the sensor.
What’s included:
flow sensor including 6-ft cable, transmitter
Specifications
Description
Ultrasonic Flowmeter
Product Type
Ultrasonic Flowmeters
Max operating temperature (º F)
140
Max operating temperature (º C)
60
Process connection
1/16" OD
Accessories
Qty
/
Price
Availability
DIN-Rail Power Supply 60W (24 VDC, 2.5 A)
EW-26901-12
at
$140.00 USD/EACH
Usually ships in 5 days.
Add to Cart
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; Liquid Flowmeter, 1 ccm to 300 LPM water
EW-17080-12
at $250.00 USD/EACH
Usually ships in 15 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
09-10 Catalog
Related technical articles (Roll over for summary)
Velocity-Profile Deviations Influence Flowmeter Performance
Selecting the Right FlowmeterPart 1
Selecting the Right FlowmeterPart 2
Show All...
Electron zap turns flimsy plastic into sturdy shrink wrap
Basics of Cartridge Filtration
Bitter Brew
Basics of Cartridge Filtration
Bitter Brew
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Job Title
|
Category
|
Brand
|
Part Number
|
Price
View product categories by:
Popularity
|
Favorites
|
Job Title
|
Price
|
Part Number
View popular items by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Job Title
View reviews by:
Popularity
|
Product Title
|
Best Rated