log in or register
Lab algicide, 8 oz bottle
EW-08796-00
$59.00 USD/EACH
  • Keeps water pure and tanks and baths clean
Prevents algae growth in baths, as well as in pumps, circulators, incubators, automatic analyzers, and blood gas analyzers. A few drops per gallon keeps water pure and odor-free. One 8-oz bottle treats up to 200 gallons.
Product Type Algicide
Product Type Algicide
Brand PolyScience
Manufacturer number 004-300040
Technical Resources
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title