Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

Labconco Fiberglass 30 fume hood, no blower, 115V

EW-09066-10
$4,264.00 USD/EACH

Call 1-888-358-4705 for availability.
Qty

  • Small fume hoods with big benefits
These 30-inch fiberglass fume hoods save space while still offering many of the features and benefits of the larger hoods. Use these efficient hoods for applications requiring small work space and good fume removal.

One-piece fiberglass interior gives this hood outstanding chemical- and fire-resistance. Pre-set baffle and by-pass airflow design assure consistent ventilation regardless of sash position. The steel-constructed exterior is glacier white epoxy-coated. Hoods feature a vertical-rising tempered glass safety sash with epoxy-coated aluminum handle.

Available with or without a built-in blower, in standard or spark-proof configurations. Hoods requires a work surface—order separately. Complete your fume hood with a variety of accessories, including a storage cabinet and an air monitor (order separately). Call our Applications Specialists regarding blower sizing and ordering service fixtures, electrical duplexes, and other custom features.

What’s included:All models are shipped fully assembled. Hard-wired installations.
Product Type Ducted Fume Hood
Interior dimensions (" W) 25
Interior dimensions (" H) 36
Interior dimensions (" D) 22.625
Exterior Dimensions (" W) 30
Exterior Dimensions (" H) 48
Exterior Dimensions (" D) 30.125
Power (VAC) 115
CE Compliance Yes
Brand Labconco

Accessories
Qty/Price
Availability

at $739.00 USD/EACH

Call 1-888-358-4705 for availability.

at $1,256.00 USD/EACH

Call 1-888-358-4705 for availability.

at $2,676.00 USD/EACH

Call 1-888-358-4705 for availability.

at $1,502.00 USD/EACH

Call 1-888-358-4705 for availability.

at $908.00 USD/EACH

Call 1-888-358-4705 for availability.




Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Leasing Available
    Financing is avaliable for many of our products.

    See Details
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title