- Identify your lab samples quickly and legibly
- One-handed operation with large display and backlight
- Advanced serialization, barcoding and symbols
- Cuts and holds printed labels until you need them
The BMP 21 portable label printer is an all-in-one labeler. With label materials designed for a variety of environments, the BMP 21 portable printer can quickly and easily create clear, legible labels that stay in place for yearsdespite extreme temperatures and curved or highly textured surfaces. Printer operates on either six AA batteries or AC power adapter (neither included; order separately).
Print labels in sizes specifically made for vials and slides. Vial size fonts are preprogrammed for 0.6, 1.5, and 2.0 mL. The laboratory-specific labels resist most chemicals, including alcohol, ethanol, and the solvent DMSO. Choose from 3/8", 1/2", and 3/4" wide labels in white or clear tape. Labels are designed for normal to extreme conditions, like liquid nitrogen or hot water
baths. To choose the correct label for your application, refer to the special properties column in the table at right.
What's Included:
one 3/4" (1.9 cm) white polylabel cartridge.