United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 3
of
13 >
Representative photo only
Laboratory Hydrogen Generator 600mL min 115 230 VAC
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-86440-03
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Life Science
>
Chromatography Products
>
Gas Chromatography (GC)
>
Gas Generators
>
Laboratory Gas Generators for GC Applications
Share
|
Laboratory Hydrogen Generator; 600mL/min; 115/230 VAC
EW-86440-03
$9,900.00 USD/EACH
Usually ships in 10 days.
Qty
:
Add to Cart
Save to shopping list
Modular and stackable units of hydrogen, nitrogen, and air gas generatorsall controlled through one CPUprovide you the flexibility your lab needs!
Assemble in any combinationeach delivers a different gas for your analytical application
CPU for control up to 32 unitscan be remotely monitored and controlled via Ethernet/Internet
Integral operating log with maintenance tables, report alarms
Stack vertically to reduce footprint and save valuable lab space
Hydrogen Generators
Reliable, uninterrupted production, no downtime to change cylinders. Continuous output of >99.9995% pure hydrogencan be used to supply both detectors and to supply carrier gas.
Integrated safety systems, which can be interfaced with the CPU or PCs for remote control
Removable water feed tank
Nitrogen Generators
capable of delivering 500 mL/min with purity > 99.9995%
available in a standard version and the HC version which ensures 100% removal of hydrocarbons (<0.1 ppm for applications with high-sensitivity detectors)
Ability to integrate with dedicated air compressor
Zero Air Generators
capable of supplying 3,000 m>/min, complete with dryer and catalytic reactor for hydrocarbon removal <0.1 ppm
high-efficiency Palladium-Platinum HC system for removal of HCTs < 0.1 ppm for an absolutely
stable and disturbance-free baseline
Serves up to 8 FIDs/NPDs
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Sourcing Hydrogen for Gas Chromatography
Chromatography Column and Phase Cross-Reference
Common Chromatography Terms
Show All...
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title