Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

Laboratory Safety Spill Kit; 3.5 Gal.; 1 Pack/case

EW-86295-07
$155.00 USD/EACH

  • Contains everything needed to safely and easily remove hazardous spills
Laboratory Safety Spill Kit neutralizes organic and inorganic acids, hydrofluoric acid, caustics-bases, halogens and solvents, as well as vapors and odors. Neutralizing agent converts hazardous chemicals to safe, non-toxic organic salts that can be easily disposed of, eliminating disposal costs and safety issues. PH indicators eliminate need for pH meters or strips.

What's Included:
1 Qt. Ultimate Acid Eater, 1 Qt. Base Eater, 1 Qt. HF Acid Eater, 1 Qt. BioRem-2000 Surface Cleaner, 1 Qt. Halex, Goggles, 8 mil vinyl aprons, 18 mil latex gloves, industrial sponge, sprayer applicator
Description LABORATORY SAFETY SPILL KIT
Qty/cs 1 PACK/ CASE
Quantity 3.5 GAL.
Manufacturer number 3500-035
Model 3500-035





Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title