United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
15 >
EW-70071-00
Leeson NEMA Type 56C-face TEFC motor, 1/3 hp, 1750 rpm, 90 VDC
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-70071-00
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Pumps
>
Motors
>
DC Motors
>
Leeson NEMA Type 56C-Face DC Motors
Share
|
Leeson NEMA Type 56C-face TEFC motor, 1/3 hp, 1750 rpm, 90 VDC
EW-70071-00
$359.00 USD/EACH
Usually ships in 4 days.
Qty
:
Add to Cart
Save to shopping list
Low-cost DC motors available in numerous configurations to meet your needs
Power pulleys, belts, pumps, or other machinery
Operate low-voltage models (12 or 24 VDC) with battery or solar power for fixed-speed applications
Explosion-proof models are Class 1, Groups C & D; Class II, Groups F & G rated
DC motors are suited for a variety of environments.
Totally enclosed, fan-cooled (TEFC) motors
utilize an external fan to circulate outside air to the motor for applications where the motor will be running continuously.
Totally enclosed, nonvented (TENV) motors
are designed without an air opening to protect against dust and other debris.
Explosion-proof (XPRF) motors
are approved for areas where hazardous materials are used.
Washdown (WDN) motors
are suitable for applications requiring regular washing or sanitizing cycles.
To vary the speed on a DC motor, order a DC controller at the side.
Specifications
Product Type
NEMA DC motors
hp
1/3
Speed range
1750 rpm max
Shaft dia
5/8"
Shaft centerline
3-1/4"
Power
90 VDC
Frame size
56C
Dimensions
11-1/2"L x 8"W x 6-1/2"H
Manufacturer number
098004
Model
098004
Brand
Leeson
Accessories
Qty
/
Price
Availability
Advanced Variable-Speed DC Motor Drive, for 1/4 to 2 hp motors
EW-70100-00
at
$689.00 USD/EACH
Usually ships in 4 days.
Add to Cart
Basic Variable-Speed DC Motor Controller for 1/4 to 2 hp motors
EW-70100-10
at
$305.00 USD/EACH
Usually ships in 4 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Motor Enclosure Terminology
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title