United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 5
of
6 >
EW-50323-20
Lightnin
®
high-torque direct-drive mixer, variable-speed, offset clamp, 115 VAC
Click to enlarge
More Product Details
Specifications
Calibrations
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-50323-20
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17050-03
Innocal NIST Calibration for Mixer
$115.00 USD/EACH/EACH
Usually ships in 13 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Lab Equipment
>
Mixers and Blenders
>
Industrial Mixers
>
Batch Mixers
>
Lightnin Batch Industrial Mixers
>
Lightnin High-Torque, Direct-Drive Industrial Mixers
Share
|
Lightnin high-torque direct-drive mixer, variable-speed, offset clamp, 115 VAC
EW-50323-20
$2,084.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Large selection lets you match mixer capability to your specific application
Mix up to 200 gallons of fluid!
These mixers offer more horsepower and torque than the small batch mixers.
High-torque mixers are available with either 20° offset tank clamps or 2" (51 mm) NPT(M) bung adapters (models 50323-12 and -22). The offset clamp positions mixer for angular, off-center mounting on unbaffled tank rims. Bung-entering models mount onto 2" (50.8 mm) NPT(F) bungs for closed-container mixing, and come with Lipseal.
What's Included:
316 stainless steel (SS) shaft, 316 SS A-310 impeller, and a drive quill with set screw.
Specifications
Product Type
Mixer
Impeller size
3.8 in
Min speed (rpm)
90
Max speed (rpm)
1800
Motor (hp)
0.25
Motor type
TENV
Mounting type
offset clamp
Shaft dia
0.625 in
Shaft length
48 in
Max torque (in-oz)
140
Power (VAC)
115
Brand
Lightnin
Calibrations
Qty
/
Price
Availability
Innocal NIST Calibration for Mixer
EW-17050-03
at $115.00 USD/EACH
Usually ships in 13 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Smart Mixing
Mixer Selection Guide
Positioning a Mixer in a Cylindrical Vessel
Show All...
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title