United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
Representative photo only
Liquid Nitrogen withdrawal system
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Instruction manual
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-03773-67
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Cooling Equipment
>
Cryogenics
>
Cryogenic Storage Dewars
>
Taylor Wharton Cryogenic Storage Dewars
>
Taylor-Wharton Liquid Nitrogen Withdrawal System
Share
|
Liquid Nitrogen withdrawal system
EW-03773-67
$778.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Permits safe and easy filling, venting, and transferring of liquid nitrogen
Dispenses liquid nitrogen at the rate of 8 L/min at 10 psi. Unique design lets you fill the storage dewar without removing the withdrawal system or venting system pressure. For storage dewars 03773-57, -59, and -61.The liquid withdrawal device enables the LD25, LD35, or LD50 Dewars to conveniently pressure transfer liquid nitrogen at rates up to 8 liters per minute. The low pressure assures safe dispensing through a phase separator or an accessory transfer hose.
Specifications
Product Type
Liquid Nitrogen Withdrawl Systems
Brand
Taylor-Wharton
Accessories
Qty
/
Price
Availability
Stainless steel cryogenic transfer hose, 4-ft long
EW-03773-40
at
$215.00 USD/EACH
In stock.
Add to Cart
Stainless steel cryogenic transfer hose, 6-ft long
EW-03773-42
at
$260.00 USD/EACH
In stock.
Add to Cart
Technical Resources
Instruction Manual
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Isolating Genomic DNA from Whole Blood
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title