United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 4
of
7 >
EW-81150-08
Brix Refractometers
Click to enlarge
More Product Details
Specifications
Calibrations
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-81150-08
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17105-00
NIST-Traceable Calibration; Refractometer
$75.00 USD/EACH/EACH
Usually ships in 9 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Lab Equipment
>
Refractometers
>
Handheld Manual Refractometers
>
Brix Refractometers
Share
|
Low-cost Brix refractometer with ATC, 0 to 32% Brix
EW-81150-08
$126.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Durable units provide quick and easy measurements
Use these economical refractometers to take quick and easy measurements. Refractometers feature adjustable focus and a zero adjustment screw for calibration. Each refractometer is calibrated and adjusted to read at 68°F (20°C). Models with automatic temperature compensation (ATC) are ideal for applications where the temperatures of your sample vary.
Refractometers are made of corrosion-resistant brass and have rubber-insulated grips. Include hard case, calibration screwdriver, and instructions.
Brix Refractometers
measure soluble solid concentration of sugars, salts, proteins, and more in foods and liquids - ideal for quality control and R&D applications. Use to measure percent Brix of juices, beverages, and other sugar-based liquids.
Specifications
Product Type
Refractometer (Analog)
Brix scale range (Minimum)
0
Brix scale range (Maximum)
32
Resolution
0.20%
Temp compensation
50 to 86°F ( 10 to 30°C)
Accuracy
±0.2%
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; Refractometer
EW-17105-00
at $75.00 USD/EACH
Usually ships in 9 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Testing Refractive Index of Candy
Refractometers
Brix Scale (sucrose)
Show All...
Equipping a Laboratory: A Study in Customer Service Excellence
Electron zap turns flimsy plastic into sturdy shrink wrap
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title