log in or register
Low density classifier assembly
EW-02658-76
$7,421.00 USD/EACH
Call 1-888-358-4705 for availability.
Qty
    Low-Density Classifier Assembly lets you take a representative sample in seconds without interrupting the drying process. Separates particles of your sample by size, shape, and moisture density. Selectively remove particles from the sample by adjusting the air flow rate - the first particles to dry will separate first since they are less dense. Use the low-density assembly for light particles like organic powders and pharmaceuticals.
    Description Low density classifier
    Accessories
    Qty/Price
    Availability
    at $264.00 USD/EACH
    Call 1-888-358-4705 for availability.
    at $41.00 USD/EACH
    Call 1-888-358-4705 for availability.
    Build a complete system
    Technical Resources
    Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Leasing Available
    Financing is avaliable for many of our products.
    See Details
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title