United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 7
of
24 >
EW-17775-30
Luminox
®
Cleaning Detergent, case of four 1-gallon bottles
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
MSDS
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-17775-30
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Laboratory Cleaning Supplies
>
Cleaning Detergents
>
Alconox Cleaning Detergents
Share
|
Luminox Cleaning Detergent, case of four 1-gallon bottles
EW-17775-30
$240.00 USD/CASE of 4
In stock.
Qty
:
Add to Cart
Save to shopping list
Order new bulk packages for added convenience
White powder Alconox
®
, Alcojet
®
, and Terg-A-Zyme
®
cleaners rinses off completelywith no residue. Alconox
®
cleaner is biodegradable; Alcojet
®
cleaner is low-sudsing; Terg-A-Zyme
®
cleaner combines the original Alconox
®
power with protein enzymes. Liqui-Nox
®
cleaner is biodegradableuse to remove soil, grease, blood, chemicals, and solvents. All cleaner are safe, odorless, and FDA certified.
ALCOJET, ALCONOX, LIQUI-NOX, TERG-A-ZYME-Reg TM Alconox Inc.
Specifications
Product Type
Equipment Cleaner
Format
Liquid
Volume (gal.)
1
Packaging
1 gallon, 4 per case
Uses
Manual or ultrasonic machine cleaning
Brand
Luminox
Manufacturer number
1901
Model
1901
Technical Resources
MSDS 1 (135 B)
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
ChemicalsSpecial Ordering Information
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title