Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
 

MAPA TRIonic Cleanroom Z-Grip Gloves, size 7-7.5, 12 pairs/pack

EW-40214-01
$40.00 USD/PACKAGE of 12

  • Added thickness for mixing, handling, and moving chemicals
Gloves feature a "Z" pattern to enhance grip when handling wet items. Made of natural rubber, nitrile, and neoprene. Individually packaged per pair.
Compliance: ISO Class 4
Product Type Gloves
Material Nitrile, neoprene, rubber
Size Medium
Length 14 in
Thickness (mil) 20
Color Cream
Brand Trionic
Manufacturer number 517317
Model 517317





Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title