Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
 

MFC 4150 Hart Communicator Intrinsically Safe, includes HART lead kit, batteries, communication/AC adapter dongle, RS-232 serial cable, universal power adapter, protective boot, and carrying case.

EW-23001-36
$3,765.00 USD/EACH

Call 1-888-358-4705 for availability.
Qty

  • The latest HART communicator from Meriam Process Technologies
  • The MFC offers several unique features that provide HART users with a more useful, more reliable and more available communication tool. Just consider the MFC 4150’s features:
  • Quick start up and connect: 15 seconds!
  • 60 hours battery life (w/o backlight)
  • Update new device profiles and firmware from Meriam via the Internet – the MFC 4150 is never out of your hands
  • Clone / Upload / Download configurations
  • Off-line mode for editing / reviewing configurations
  • Document and store 200 configurations
  • As-found / As-left history with Meriam DMS software
  • 13-line, 128 x 128 graphic, backlit display
  • Dongle coordinates AC power adapter and serial communications
  • Full alpha-numeric keypad
  • Review / Edit on the fly
  • Quick Menu short cuts for commonly used commands
  • HART menu Home key
  • Dedicated text edit keys
  • Programmable lock-out settings
  • Contrast key for direct adjustment in any mode
  • Elastic hand strap, adjustable shoulder strap and soft carrying case
  • Left side thumb keys for one-handed navigation
The MFC 4150 HART® Communicator is the latest HART communicator from Meriam Process Technologies. HART field devices can be configured and trimmed using the MFC. The MFC is a full function HART Communicator with features that optimize commissioning, configuration and maintenance operations.
What's Included:
DMS Software Datasheet
HART® lead kit
Protective Rubber Boot
RS-232 Serial Cable
Power adapter 120 to 240 VAC, 50 –
60 Hz and Universal Plug Adapter Kit
AA alkaline batteries (6 each)
Hard copy user’s manual
DPC Manager Utility for updating MFC
CE for EMC testing (all models)
Product Type Accessory
Display 13 line, 128 x 128 pixel, backlight
Housing material ABS plastic case
Dimensions 4.25 to 2.5 in W x 10.375 in H x 1.875 in D
Power 6 AA alkaline or NIMG batteries or 100-240 VAC power adapter
CE Compliance Yes

Accessories
Qty/Price
Availability

at $89.00 USD/EACH

Usually ships in 14 days.

at $125.00 USD/EACH

Call 1-888-358-4705 for availability.

at $18.00 USD/EACH

Usually ships in 14 days.

at $113.00 USD/EACH

Usually ships in 14 days.

at $24.00 USD/EACH

In stock.

at $17.00 USD/EACH

Usually ships in 14 days.

at $118.00 USD/EACH

Usually ships in 14 days.

at $114.00 USD/EACH

Usually ships in 14 days.




Technical Resources
 
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.

See Details
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title