United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
4 >
Representative photo only
MNOVA NMR Predict Plug In Annual License for Academia and Government NMR Software for processing analysis simulation and reporting
Click to enlarge
More Product Details
Specifications
Technical Resources
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-83090-72
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Material Testing & Physical Testing
>
Spectroscopy and Spectrophotometers
>
Nuclear Magnetic Resonance (NMR) Spectroscopy
>
MNOVA NMR Predict Desktop Plug-In for Spectrometers
Share
|
MNOVA NMR Predict Plug-In Annual License, for Academia and Government- NMR Software for processing, analysis, simulation and reporting
EW-83090-72
$140.00 USD/EACH
Usually ships in 4 days.
Qty
:
Add to Cart
Save to shopping list
Easily predict 1-D and HSQC NMR spectra from a molecular structure
Verify your experimental spectra with prediction data
Keep your Mnova software up-to-date with one year of support and updates for free
Automatic solvent recognition and intelligent peak picking ensures optimal results
MNOVA NMRPredict Desktop Plug-in
generates of 1-D and Heteronuclear Single Quantum Coherence (HSQC) spectra from molecular structures and is ideal for the expert user looking for extensive or advanced functionality. Combined with the MNOVA NMR software, it provides a very simple interface to automatically analyze spectra. Features such as peak picking, integration, and multiplet analysis allow your experiments to run smoothly. The plug-in allows you to assign 1-D and 2-D spectra, make notes and reports in various journal formats, and paste molecular structures into your document. Further automation is allowed and access to pre-made scripts and processing templates is available; contact our Application Specialists for more details. The plug-in will operate in Windows®, Mac®, and various Linus environments.
The plug-in is available with either a perpetual or annual license. The perpetual license allows you to use the licensed software indefinitely. For the first year after purchase, the user is entitled to download any updates to the software and receive a full free year of technical support. Once the initial year ends, you can continue to purchase updates and support packages. The annual license will allow one year use of the licensed software, including updates and free technical support. Once the initial twelve months ends, the software will no longer function unless a new license is purchased.
Technical Resources
Related technical articles (Roll over for summary)
Seeing the Light: An Overview of Visible and UV-VIS Spectroscopy
Spectrophotometer Selection Guide
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title