log in or register
MNOVA NMR Predict Plug-In Perpetual License, for Academia and Government- NMR Software for processing, analysis, simulation and reporting
EW-83090-73
$350.00 USD/EACH
Usually ships in 4 days.
Qty
  • Easily predict 1-D and HSQC NMR spectra from a molecular structure
  • Verify your experimental spectra with prediction data
  • Keep your Mnova software up-to-date with one year of support and updates for free
  • Automatic solvent recognition and intelligent peak picking ensures optimal results
MNOVA NMRPredict Desktop Plug-in generates of 1-D and Heteronuclear Single Quantum Coherence (HSQC) spectra from molecular structures and is ideal for the expert user looking for extensive or advanced functionality. Combined with the MNOVA NMR software, it provides a very simple interface to automatically analyze spectra. Features such as peak picking, integration, and multiplet analysis allow your experiments to run smoothly. The plug-in allows you to assign 1-D and 2-D spectra, make notes and reports in various journal formats, and paste molecular structures into your document. Further automation is allowed and access to pre-made scripts and processing templates is available; contact our Application Specialists for more details. The plug-in will operate in Windows®, Mac®, and various Linus environments.
The plug-in is available with either a perpetual or annual license. The perpetual license allows you to use the licensed software indefinitely. For the first year after purchase, the user is entitled to download any updates to the software and receive a full free year of technical support. Once the initial year ends, you can continue to purchase updates and support packages. The annual license will allow one year use of the licensed software, including updates and free technical support. Once the initial twelve months ends, the software will no longer function unless a new license is purchased.
Technical Resources
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title