log in or register
Maintenance kit for picoSpin-45 benchtop NMR compatible with 1/32" fittings
EW-83090-53
$52.50 USD/EACH
Usually ships in 11 days or sooner.
Qty
    The picoSpin-45 Cartridge Maintenance Kit provides accessory items needed to perform maintenance on the capillary inlet filter of the picoSpin-45 miniature NMR spectrometer. The kit includes 1/4” and 3/16” inch open-end box wrenches, 10 replacement 2 µm frit filters, and 2 short pieces of wire to help with the removal of frits. Also included are instructions describing how to ‘back-flush’ clean the existing frit filter and instructions on replacing a frit filter.
    In order to know if you have 1/16-inch or 1/32-inch fittings on your picoSpin, look at the inlet fitting of your picoSpin-45. If you see two stainless hex nuts, you have 1/32-inch fittings. If you see one stainless hex nut, you have 1/16-inch fittings.
    In general, capillary cartridges shipped before January 2013 have 1/32-inch fittings. Capillary cartridges shipped after January 2013 generally will have 1/16-inch fittings. Please contact our applications specialists if you need further assistance.
    Brand picoSpin
    Technical Resources
    Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title