United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
3 >
Representative photo only
Masterflex I P Easy Load pump head PSF housing CRS rotor
Click to enlarge
More Product Details
See
Upgraded
model
Specifications
Accessories
Replacement parts
Technical Resources
Instruction manual
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
SI-77601-00
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Masterflex Peristaltic Pumps
>
Masterflex Pump Heads
>
Masterflex I/P Pump Heads
>
I/P Easy-Load Pump Heads
Share
|
Masterflex I/P Easy-Load pump head, PSF housing/CRS rotor.
SI-77601-00
$510.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Quick, simple tubing changestubing replacement and fluid changeover takes only seconds
Accept multiple tubing sizes for maximum flexibility
Automatic tubing retention eliminates user adjustments and makes setup easyoffers improved tubing life and improved performance
Fast tubing changes minimize downtime
Adjustable occlusion lets you optimize pressure and suction capabilities and tubing life by adjusting the “squeeze” on the tubing.
Five-screw mounting—ensures pump head is mounted securely to drive
The Easy-Load pump head combines an over-center cam with automatic tubing retention in a rugged, chemical-resistant housing. This patented design allows fast, easy tubing changes without removing the pump head from the drive. Built-in tubing guide ensures correct positioning of tubing to maximize performance and pump tubing life. Spring-loaded tubing retainers ensure tubing is secured automatically to promote long tubing life and optimize performance of each tubing size.
Select from polysulfone (PSF) housing with CRS or SS rotor, or polyphenylene sulfide (PPS) housing with SS rotor. PSF is economical, while PPS offers enhanced chemical resistance. SS rotors feature PTFE-sealed bearings for long life and reduced maintenance; CRS rotors have Buna N shielded bearings.
Dual-channel hardware allows two pump heads to operate on a single drive—order 77601-96 separately under "Accessories".
With single or multiple channels, tubing can be changed in each channel without removing the EASY-LOAD pump heads from your drive.
What's Included:
single-channel mounting hardware and a 20" (51 cm) length of silicone pump tubing.
Specifications
Product Type
Pump Head
Max flow rate (LPM)
13.0
Max flow rate (GPM)
3.5
Min flow rate (LPM)
0.01
Min flow rate (GPM)
0.002
Accepted tubing
I/P 26, I/P 73, I/P 82
Max pressure (psi)
40
Max pressure (bar)
2.7
Rotor material
Cold-rolled steel
Housing material
PSF
Occlusion
Adjustable
Flow Chart
Click to View
Automatic tubing retention
Yes
Brand
Masterflex
Manufacturer number
77601-00
Accessories
Qty
/
Price
Availability
Stainless steel mounting hardware for two I/P Easy-Load pump heads.
EW-77601-96
at
$32.25 USD/EACH
In stock.
Add to Cart
Upgraded Model
Qty
/
Price
Availability
Masterflex I/P High-Performance Pump Head, Polyester/SS Housing with SS Rotor.
EW-77600-62
at $790.00 USD/EACH
In stock.
Add to Cart
Replacement parts
Qty
/
Price
Availability
Replacement I/P Tang Boots. Pack of 10.
EW-77600-03
at $67.50 USD/PACKAGE
In stock.
Add to Cart
Stainless steel mounting hardware for one I/P Easy-Load pump head.
EW-77601-95
at $27.25 USD/EACH
In stock.
Add to Cart
Technical Resources
Instruction Manual.pdf
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Masterflex® Pump Decreases the Risk of Heavy Metal Exposure
Masterflex: How To Load Tubing - EZ Load
I/P® Pump Head Flow Performance
Show All...
A Sweet Success: Famous Maple Syrup Producer Enhances Production with Masterflex® B/T® Variable-Speed Pump
Masterflex Connection Newsletter Vol.4-3 - Industrial Strength Pumps
Technical Expertise: How Cole-Parmer Application Specialists Respond to Customers' Needs
Reducing Pulsation
Peristaltic Tubing Pumps in Dispensing Applications
Masterflex Tubing: Questions and Answers
Masterflex Tubing Pump Systems FAQs
Tygon® Flexible Plastic Tubing to Tubing Solvent Sealing
Peristaltic Pumps Not Just for Labs Anymore
Tubing Selection Guide For Viscous Fluids
Pumping Abrasives
Pumping Viscous Fluids
Peristaltic Pumps Offer Custom Fluid Solutions
Peristaltic Pumps Offer Economical Multichannel Pumping
Reducing Pulsation in Peristaltic Pumping Systems
Pumping Abrasives
Masterflex Systems: Questions and Answers
High-Performance Masterflex Pump Head
Six Ways to Simplify Fluid Transfer
Masterflex Pump FAQs
Masterflex Tubing Formulations
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title