United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
EW-77600-62
Masterflex
®
I/P
®
High-Performance Pump Head, Polyester/SS Housing with SS Rotor.
Click to enlarge
More Product Details
Specifications
Replacement parts
Reconditioned & Like New
Technical Resources
Instruction manual
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-77600-62
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Masterflex Peristaltic Pumps
>
Masterflex Pump Heads
>
Masterflex I/P Pump Heads
>
I/P High-Performance Pump Head
Share
|
Masterflex I/P High-Performance Pump Head, Polyester/SS Housing with SS Rotor.
EW-77600-62
$790.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Highest flow rates, best pressure and suction performance of any I/P pump head
Polyester and stainless steel (SS) construction withstands tough process environments
Versatile mounting configurations—mount with tubing exiting to the top or side for flexible setup
C-shaped design allows tubing to enter and exit the same side of the pump headfacilitates connection to tubing inlet and outlet
Adjustable tubing retention; adjust to prevent tubing movement in head
Ideal for industrial process and production applications requiring a single pump head, this heavy-duty three-roller pump head produces flow rates up to 19 LPM—the highest of any I/P pump head. Simple to load, it accepts Masterflex I/P high-performance precision pump tubing for greater pressure generation and suction lift, enhanced ability to pump viscous fluids, and longer tubing life. Adjustable tubing retention holds tubing securely in place and prevents movement while in the head.
The I/P High-Performance pump heads are compatible with I/P drives that accept two or more pump heads. Drives that accept two heads have enough torque to run pump head even under difficult conditions such as high back pressure.
Note: High-Performance pump head is not stackable.
What's Included:
mounting hardware, a 20" (51 cm) length of silicone pump tubing, and replaceable tang boot for quiet operation and long service life
Specifications
Product Type
Pump Head
Max flow rate (LPM)
19.0
Max flow rate (GPM)
4.5
Min flow rate (LPM)
0.01
Min flow rate (GPM)
0.002
Accepted tubing
I/P 70, I/P 88, I/P 89
Max pressure (psi)
40
Max pressure (bar)
2.7
Rotor material
Stainless steel
Housing material
Stainless steel, thermoset polyester
Occlusion
Fixed
Flow Chart
Click to View
Brand
Masterflex
Manufacturer number
77600-62
Replacement parts
Qty
/
Price
Availability
Repl. moutning screws for 77600-62
EW-77600-01
at $19.80 USD/SET
Usually ships in 7 days.
Add to Cart
Replacement I/P Tang Boots. Pack of 10.
EW-77600-03
at $67.50 USD/PACKAGE
In stock.
Add to Cart
Repl. tubing retainer assembly, for 77600-62
EW-77600-66
at $84.00 USD/EACH
In stock.
Add to Cart
Repl. occlusion bed assembly for 77600-62
EW-77600-67
at $119.00 USD/EACH
In stock.
Add to Cart
Rotor/body kit
EW-77600-69
at $542.00 USD/EACH
Usually ships in 7 days.
Add to Cart
Reconditioned & Like New
Qty
/
Price
Availability
RECOND 77600-62
EW
-
00151SX
at
$414.00 USD/EACH
In stock.
Add to Cart
Technical Resources
Instruction Manual (286 KB)
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Masterflex: How To Load Tubing - High Performance Pumps
I/P® Pump Head Flow Performance
A Sweet Success: Famous Maple Syrup Producer Enhances Production with Masterflex® B/T® Variable-Speed Pump
Show All...
Masterflex Connection Newsletter Vol.4-3 - Industrial Strength Pumps
Technical Expertise: How Cole-Parmer Application Specialists Respond to Customers' Needs
Reducing Pulsation
Peristaltic Tubing Pumps in Dispensing Applications
Masterflex Tubing: Questions and Answers
Masterflex Tubing Pump Systems FAQs
Tygon® Flexible Plastic Tubing to Tubing Solvent Sealing
Peristaltic Pumps Not Just for Labs Anymore
Tubing Selection Guide For Viscous Fluids
Pumping Abrasives
Pumping Viscous Fluids
Peristaltic Pumps Offer Custom Fluid Solutions
Peristaltic Pumps Offer Economical Multichannel Pumping
Reducing Pulsation in Peristaltic Pumping Systems
Pumping Abrasives
Masterflex Systems: Questions and Answers
High-Performance Masterflex Pump Head
Six Ways to Simplify Fluid Transfer
Masterflex Pump FAQs
Masterflex Tubing Formulations
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title