United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 3
of
5 >
Representative photo only
Masterflex I P NEMA Type 56 C pump head adapter 9 7 1 adapter gear ratio 650 rpm maximum pump head speed ATEX Zone 2 approved
Click to enlarge
More Product Details
Specifications
Technical Resources
Instruction manual
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-77490-20
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Masterflex Peristaltic Pumps
>
Masterflex Pump Head Adapters
>
I/P Pump Head Adapters
>
Masterflex I/P Pump Head Adapters
Share
|
Masterflex I/P NEMA Type 56 C pump head adapter, 9.7:1 adapter gear ratio, 650 rpm maximum pump head speed, ATEX Zone 2 approved
EW-77490-20
$618.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Flow range: 0.12 to 13 LPM (0.03 to 3.5 GPM). Flow rate depends on motor rpm, adapter gear ratio, and tubing size. Pump head range is 20 to 650 rpm.
Use one or two I/P pump heads. Pump two different fluids simultaneously at the same or different flow rates.
Cast-aluminum adapters feature built-in gear box and are IP56 rated—includes mounting hardware. Adapters maintain integrity of your motor rating up to IP56.
Adapters accept two different pump head types: Standard and EASY-LOAD
®
Use these adapters to mount MASTERFLEX I/P pump heads to 56C frame motors or to a metric IEC 72/ISO 63 motor with B5 flange.
Use the following formula to calculate flow rate— do not exceed 650 rpm pump head speed: Flow rate = (Motor rpm/adapter gear ratio) x flow per revolution
Specifications
Motor mount
NEMA Type 56C
Gear ratio
9:7:1
Drive requirements
1/10 hp to 1/2 hp; 650 rpm pump head speed max
Mounting type
Direct coupling
Dimensions
6-3/4"L x 6-1/2"W x 6-1/2"H (17.2 cm x 16.5 cm x 16.5 cm)
Brand
Masterflex
Manufacturer number
77490-20
Technical Resources
Instruction Manual (124 KB)
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Technical Expertise: How Cole-Parmer Application Specialists Respond to Customers' Needs
Reducing Pulsation
Peristaltic Tubing Pumps in Dispensing Applications
Show All...
Masterflex Tubing: Questions and Answers
Masterflex Tubing Pump Systems FAQs
Tygon® Flexible Plastic Tubing to Tubing Solvent Sealing
Peristaltic Pumps Not Just for Labs Anymore
Tubing Selection Guide For Viscous Fluids
Pumping Abrasives
Pumping Viscous Fluids
Peristaltic Pumps Offer Custom Fluid Solutions
Peristaltic Pumps Offer Economical Multichannel Pumping
Reducing Pulsation in Peristaltic Pumping Systems
Pumping Abrasives
Masterflex Systems: Questions and Answers
High-Performance Masterflex Pump Head
Six Ways to Simplify Fluid Transfer
Masterflex Pump FAQs
Masterflex Tubing Formulations
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title