United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 2
of
2
EW-77963-20
Masterflex
®
I/P
®
precision brushless pump system with High-Performance pump head 77600-62, 115/230 VAC.
Click to enlarge
More Product Details
Specifications
Calibrations
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-77963-20
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17050-01
NIST-Traceable Calibration for Peristaltic Pump Drive
$115.00 USD/EACH/EACH
Usually ships in 8 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Masterflex Peristaltic Pumps
>
Masterflex I/P Tubing Pumps
>
I/P Complete Pump Systems
>
I/P Analog Pump Systems
>
I/P Precision Brushless Process Pumps
Share
|
Masterflex I/P precision brushless pump system with High-Performance pump head 77600-62, 115/230 VAC.
EW-77963-20
$2,855.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Flow: 0.7 to 17 LPM (0.2 to 4.5 GPM) with included I/P
®
88 tubing; flow rate depends on motor rpm and tubing size.
Three-turn speed control offers precision and repeatabilityLED display shows percent speed with 1% resolution
1/3-hp brushless motor handles challenging applications with minimal maintenance
Non-chip epoxy-coated, IP55-rated steel and aluminum enclosure protects against moisture, chemicals, and dust
Compact drives are light enough to carry with one hand, but powerful enough to drive two pump heads
Specifications
Product Type
Peristaltic Pump
Min flow rate (LPM)
0.7 LPM (0.2 GPM)
Max flow rate (LPM)
17.0
Tubing
Tygon® LFL I/P 88
06429-88
Drive
I/P brushless process drive 77410-10
Pump head
I/P High-Performance pump head 77600-62
rpm
33 to 650
Power (VAC)
90 to 260
Power (Hz)
50/60
Power (Amps)
3.5 at 115 VAC
1.8 at 230 VAC
Manufacturer number
77963-20
Pump Series
I/P
Brand
Masterflex
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration for Peristaltic Pump Drive
EW-17050-01
at $115.00 USD/EACH
Usually ships in 8 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
A Sweet Success: Famous Maple Syrup Producer Enhances Production with Masterflex® B/T® Variable-Speed Pump
Masterflex Connection Newsletter Vol.4-3 - Industrial Strength Pumps
Technical Expertise: How Cole-Parmer Application Specialists Respond to Customers' Needs
Show All...
Reducing Pulsation
Peristaltic Tubing Pumps in Dispensing Applications
Masterflex Tubing: Questions and Answers
Masterflex Tubing Pump Systems FAQs
Tygon® Flexible Plastic Tubing to Tubing Solvent Sealing
Peristaltic Pumps Not Just for Labs Anymore
Tubing Selection Guide For Viscous Fluids
Pumping Abrasives
Pumping Viscous Fluids
Peristaltic Pumps Offer Custom Fluid Solutions
Peristaltic Pumps Offer Economical Multichannel Pumping
Reducing Pulsation in Peristaltic Pumping Systems
Pumping Abrasives
Masterflex Systems: Questions and Answers
High-Performance Masterflex Pump Head
Six Ways to Simplify Fluid Transfer
Masterflex Pump FAQs
Masterflex Tubing Formulations
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title