log in or register
Masterflex L/S Easy-Load II pump head for high-performance precision tubing, fixed occlusion, CRS rotor.
SI-77200-52
$228.00 USD/EACH
  • Automatic retention holds tubing securely in place with no manual adjustment—just load the tubing and start the pump
  • Four-roller rotor improves pressure performance and mechanical stability while reducing pulsation.
  • Improved occlusion bed geometry lengthens tubing life.
  • Polyphenylene sulfide (PPS) housing with either stainless steel (SS) or cold-rolled steel (CRS) rotor—choose CRS for economy or SS for greater durability and chemical resistance.
  • Model 77201-62 with adjustable occlusion gives you the option of loosening occlusion for longer tubing life or increasing occlusion for higher pressure.
  • Each pump head for high-performance precision pump tubing accepts tubing sizes L/S® 15, L/S® 24, L/S® 35, and L/S® 36. Achieve flow rates of 1.7 to 2900 mL/min (1 to 600 rpm).
These pump heads accept the four high-performance precision tubing sizes. High-performance precision tubing has a thicker wall than precision tubing enabling better pressure generation, greater suction lift, and greater ability to pump viscous fluids.
What's included: single-channel mounting hardware and a 15'' (38-cm) length of silicone pump tubing. For stacking multiple heads (up to the specified limits of your drive) order two- three- or four-channel hardware separately.
Hot Tips:
With single or multiple channels, you can change tubing in each channel without removing the EASY-LOAD II pump head(s) from your drive.
Product Type Pump Head
mL/rev Depends on tubing selected.
Accepted tubing L/S® 15, L/S 24, L/S 35, L/S 36
Rotor material Cold-rolled steel
Housing material Polyphenylene sulfide
Pressure Chart Click to View
Rollers Four; cold-rolled steel
Occlusion Fixed
Dimensions 5"W x 4-3/4"H x 2-3/4"D
(12.7 x 12.1 x 7.0 cm)
Flow Chart Click to View
Automatic tubing retention Yes
Brand Masterflex
Manufacturer number 77200-52
Accessories
Qty/Price
Availability
at $34.75 USD/SET of 1
In stock.
at $44.75 USD/SET of 1
In stock.
at $49.00 USD/SET of 1
In stock.
Upgraded Model
Qty/Price
Availability
at $368.00 USD/EACH
In stock.
Replacement parts
Qty/Price
Availability
at $22.75 USD/SET of 1
In stock.
Technical Resources
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title