United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 3
of
3
Representative photo only
Masterflex L S Easy Load II pump head for precision tubing adjustable occlusion SS rotor
Click to enlarge
More Product Details
See
Upgraded
model
Specifications
Accessories
Replacement parts
Technical Resources
Instruction manual
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Related media
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-77201-60
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Masterflex Peristaltic Pumps
>
Masterflex Pump Heads
>
Masterflex L/S Pump Heads
>
L/S Easy-Load II Pump Heads
>
Masterflex L/S Easy-Load II Pump Heads for Precision Tubing
Share
|
Masterflex L/S Easy-Load II pump head for precision tubing, adjustable occlusion, SS rotor.
EW-77201-60
$338.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Automatic retention holds tubing securely in place with no manual adjustmentjust load the tubing and start the pump
Four-roller rotor improves pressure performance and mechanical stability while reducing pulsation.
Improved occlusion bed geometry lengthens tubing life.
Polyphenylene sulfide (PPS) housing with either stainless steel (SS) or cold-rolled steel (CRS) rotorchoose CRS for economy or SS for greater durability and chemical resistance.
Model 77201-60 with adjustable occlusion gives you the option of loosening occlusion for longer tubing life or increasing occlusion for higher pressure.
Each pump head for precision pump tubing accepts tubing sizes L/S
®
13, L/S
®
14, L/S
®
16, L/S
®
25, L/S
®
17, and L/S
®
18. Achieve flow rates of 0.06 to 2300 mL/min (1 to 600 rpm).
For stacking multiple heads (up to the specified limits of your drive) order two- three- or four-channel hardware separately under "Accessories". When two Easy-Load II pump heads are stacked rollers are offset by 45°combine the outlets from two stacked heads for a 95% reduction in flow pulsation.
What's Included:
single-channel mounting hardware and a 15" (38-cm) length of silicone pump tubing.
Hot Tips:
With single or multiple channels, you can change tubing in each channel without removing the EASY-LOAD II pump head(s) from your drive.
Specifications
Product Type
Pump Head
mL/rev
Depends on tubing selected.
Accepted tubing
L/S® 13, L/S 14, L/S 16, L/S 25, L/S 17, L/S 18
Rotor material
Stainless steel
Housing material
Polyphenylene sulfide
Pressure Chart
Click to View
Rollers
Four; stainless steel
Occlusion
Adjustable
Dimensions
5"W x 4-3/4"H x 2-3/4"D
(12.7 x 12.1 x 7.0 cm)
Flow Chart
Click to View
Brand
Masterflex
Manufacturer number
77201-60
Accessories
Qty
/
Price
Availability
Stainless steel mounting hardware; for two L/S Easy-Load II pump heads.
EW-77200-02
at
$34.75 USD/SET of 1
In stock.
Add to Cart
Stainless steel mounting hardware; for three L/S Easy-Load II pump heads.
EW-77200-03
at
$44.75 USD/SET of 1
In stock.
Add to Cart
Stainless steel mounting hardware; for four L/S Easy-Load II pump heads.
EW-77200-04
at
$49.00 USD/SET of 1
In stock.
Add to Cart
Upgraded Model
Qty
/
Price
Availability
Masterflex L/S High-Performance pump head.
EW-77250-62
at $368.00 USD/EACH
In stock.
Add to Cart
Replacement parts
Qty
/
Price
Availability
Stainless steel mounting hardware; for one L/S Easy-Load II pump head.
EW-77200-01
at $22.75 USD/SET of 1
In stock.
Add to Cart
Technical Resources
Instruction Manual
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Masterflex® and the Human Heart: How One Pump is Supporting Research for the Other
Pump Head Torque Requirements
Pump Head Torque Requirements
Show All...
Occlusion: Questions and Answers
L/S® Pump Head Flow Performance
A Sweet Success: Famous Maple Syrup Producer Enhances Production with Masterflex® B/T® Variable-Speed Pump
Technical Expertise: How Cole-Parmer Application Specialists Respond to Customers' Needs
Reducing Pulsation
Peristaltic Tubing Pumps in Dispensing Applications
Masterflex Tubing: Questions and Answers
Masterflex Tubing Pump Systems FAQs
Tygon® Flexible Plastic Tubing to Tubing Solvent Sealing
Peristaltic Pumps Not Just for Labs Anymore
Tubing Selection Guide For Viscous Fluids
Pumping Abrasives
Pumping Viscous Fluids
Peristaltic Pumps Offer Custom Fluid Solutions
Peristaltic Pumps Offer Economical Multichannel Pumping
Reducing Pulsation in Peristaltic Pumping Systems
Pumping Abrasives
Masterflex Systems: Questions and Answers
High-Performance Masterflex Pump Head
Six Ways to Simplify Fluid Transfer
Masterflex Pump FAQs
Masterflex Tubing Formulations
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Related media
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title