United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
3 >
Representative photo only
Masterflex L S Easy Load pump head for precision tubing PSF housing CRS rotor
Click to enlarge
More Product Details
See
Upgraded
model
Specifications
Accessories
Replacement parts
Technical Resources
Instruction manual
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
SI-07518-00
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Masterflex Peristaltic Pumps
>
Masterflex Pump Heads
>
Masterflex L/S Pump Heads
>
L/S Easy-Load Pump Heads
>
L/S Easy-Load Pump Heads for Precision Tubing
Share
|
Masterflex L/S Easy-Load pump head for precision tubing; PSF housing, CRS rotor.
SI-07518-00
$206.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Ideal for fast tubing changes and reduced maintenance
Patented design and adjustable tubing retention allow for multiple tubing sizeschange tubing without removing head from drive.
Polysulfone (PSF) housing with either stainless steel (SS) or cold-rolled steel (CRS) rotorchoose CRS for economy or SS for greater durability and chemical resistance.
Model 07518-60 with polyphenylene sulfide (PPS) housing and SS rotor offers enhanced chemical resistance.
Each pump head for precision pump tubing accepts tubing sizes L/S
®
13, L/S
®
14, L/S
®
16, L/S
®
25, L/S
®
17, and L/S
®
18. Achieve flow rates of 0.06 to 2300 mL/min (1 to 600 rpm).
What''s included:
single-channel mounting hardware and a 15" (38-cm) length of silicone pump tubing.
Two-, three-, and four-channel mounting hardware is available separately. Stack pump heads up to the specified limits of your L/S
®
drive.
Hot Tips:
With single or multiple channels, you can change tubing in each channel without removing the EASY-LOAD pump head(s) from your drive!
Specifications
Product Type
Pump Head
mL/rev
Depends on tubing selected.
Accepted tubing
L/S® 13, L/S 14, L/S 16, L/S 25, L/S 17, L/S 18
Rotor material
Cold-rolled steel
Housing material
Polysulfone
Rollers
Three; cold-rolled steel
Occlusion
Average, fixed
Dimensions
4-1/2"W x 4"H x 2-1/2"D
(11.4 x 10.2 x 6.4 cm)
Flow Chart
Click to View
Brand
Masterflex
Manufacturer number
07518-00
Accessories
Qty
/
Price
Availability
Stainless steel mounting hardware; mounts two (2) Masterflex L/S Easy-Load pump heads.
EW-07013-05
at
$34.50 USD/SET of 1
In stock.
Add to Cart
Stainless steel mounting hardware; mounts three (3) Masterflex L/S Easy-Load pump heads.
EW-07013-08
at
$37.00 USD/SET of 1
In stock.
Add to Cart
Stainless steel mounting hardware; mounts four (4) Masterflex L/S Easy-Load pump heads.
EW-07013-09
at
$41.50 USD/SET of 1
In stock.
Add to Cart
Upgraded Model
Qty
/
Price
Availability
Masterflex L/S Easy-Load 3 pump head for precision tubing, CRS rotor.
EW-77800-50
at $205.00 USD/EACH
Usually ships in 7 days or sooner.
Add to Cart
Replacement parts
Qty
/
Price
Availability
Stainless steel mounting hardware; mounts one (1) Masterflex L/S Easy-Load pump head.
EW-07013-04
at $30.50 USD/SET of 1
In stock.
Add to Cart
Technical Resources
Instruction Manual.pdf
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Masterflex: How To Load Tubing - EZ Load
Masterflex® and the Human Heart: How One Pump is Supporting Research for the Other
Pump Head Torque Requirements
Show All...
Pump Head Torque Requirements
Occlusion: Questions and Answers
L/S® Pump Head Flow Performance
A Sweet Success: Famous Maple Syrup Producer Enhances Production with Masterflex® B/T® Variable-Speed Pump
Technical Expertise: How Cole-Parmer Application Specialists Respond to Customers' Needs
Reducing Pulsation
Peristaltic Tubing Pumps in Dispensing Applications
Masterflex Tubing: Questions and Answers
Masterflex Tubing Pump Systems FAQs
Tygon® Flexible Plastic Tubing to Tubing Solvent Sealing
Peristaltic Pumps Not Just for Labs Anymore
Tubing Selection Guide For Viscous Fluids
Pumping Abrasives
Pumping Viscous Fluids
Peristaltic Pumps Offer Custom Fluid Solutions
Peristaltic Pumps Offer Economical Multichannel Pumping
Reducing Pulsation in Peristaltic Pumping Systems
Pumping Abrasives
Masterflex Systems: Questions and Answers
High-Performance Masterflex Pump Head
Six Ways to Simplify Fluid Transfer
Masterflex Pump FAQs
Masterflex Tubing Formulations
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title