log in or register
Masterflex L/S Easy-Load pump head for precision tubing; PSF housing, SS rotor.
SI-07518-10
$258.00 USD/EACH
  • Ideal for fast tubing changes and reduced maintenance
  • Patented design and adjustable tubing retention allow for multiple tubing sizes—change tubing without removing head from drive.
  • Polysulfone (PSF) housing with either stainless steel (SS) or cold-rolled steel (CRS) rotor—choose CRS for economy or SS for greater durability and chemical resistance.
  • Model 07518-60 with polyphenylene sulfide (PPS) housing and SS rotor offers enhanced chemical resistance.
  • Each pump head for precision pump tubing accepts tubing sizes L/S® 13, L/S® 14, L/S® 16, L/S® 25, L/S® 17, and L/S® 18. Achieve flow rates of 0.06 to 2300 mL/min (1 to 600 rpm).
What''s included: single-channel mounting hardware and a 15" (38-cm) length of silicone pump tubing.
Two-, three-, and four-channel mounting hardware is available separately. Stack pump heads up to the specified limits of your L/S® drive.
Hot Tips:
With single or multiple channels, you can change tubing in each channel without removing the EASY-LOAD pump head(s) from your drive!
Product Type Pump Head
mL/rev Depends on tubing selected.
Accepted tubing L/S® 13, L/S 14, L/S 16, L/S 25, L/S 17, L/S 18
Rotor material Stainless steel
Housing material Polysulfone
Pressure Chart Click to View
Rollers Three; stainless steel
Occlusion Average, fixed
Dimensions 4-1/2"W x 4"H x 2-1/2"D
(11.4 x 10.2 x 6.4 cm)
Flow Chart Click to View
Brand Masterflex
Manufacturer number 07518-10
Accessories
Qty/Price
Availability
at $34.50 USD/SET of 1
In stock.
at $37.00 USD/SET of 1
In stock.
at $41.50 USD/SET of 1
In stock.
Upgraded Model
Qty/Price
Availability
at $256.00 USD/EACH
In stock.
Replacement parts
Qty/Price
Availability
at $30.50 USD/SET of 1
In stock.
Reconditioned & Like New
Qty/Price
Availability
RECOND 07518-10
EW-00085ZH
at $117.00 USD/EACH
In stock.
Technical Resources
  • Instruction Manual.pdf
  • Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    Customers Who Bought This Item Also Bought
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title