United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 2
of
3 >
Representative photo only
Masterflex L S Precision Variable Speed Drive with Remote Input 3 to 300 rpm 90 to 260 VAC
Click to enlarge
More Product Details
Specifications
Calibrations
Technical Resources
Instruction manual
Virtual Catalog (PDFs)
More info
Brochure
Related technical articles
Service & Support
Related media
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-07528-20
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17050-01
NIST-Traceable Calibration for Peristaltic Pump Drive
$115.00 USD/EACH/EACH
Usually ships in 8 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Masterflex Peristaltic Pumps
>
Masterflex L/S Tubing Pumps
>
Masterflex L/S Drives
>
L/S Analog Variable-Speed Drives
>
L/S Precision Variable-Speed Console Drives
Share
|
Masterflex L/S Precision Variable-Speed Drive with Remote Input; 3 to 300 rpm, 90 to 260 VAC.
EW-07528-20
$1,120.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Easy-to-use drives offer precise speed control and digital display of rpm—ideal for metering applications
Flow rate 0.06 to 3400 mL/min (depends on drive rpm and tubing size)
Three digit LED display shows motor rpmconfirm speed setting at a glance
Reversible motor—purge tubing before or after pumping; pump fluid in either direction
LED indicators confirm motor direction
Powerful motors let you stack 2 or 4 pump heads for increased flow rates and for multichannel applications
Separate speed and power (on/off) controlmaintain speed setting while turning pump on/off
Stackable, splash-resistant ABS plastic housing; IP33 rated
Remote control capabilities via a DB9 female connector on back of drive
Speed control input: 4 to 20 mA or 0 to 10 VDC
Pump direction (CW/CCW) via open collector or contact closure
Start/stop via open collector or contact closure
Membrane keypad for easy up-front control of speed, direction, and start/stop
These accurate analog console drives have powerful motors, speed display, and ±0.25% speed control for precise flow regulation—ideal for metering applications. Remote speed control requires DB9 male connector 07595-45 or cable assembly 07595-47. Use remote connector for control of start/stop via contact closure or open collector (5 V TTL); or control start/stop via footswitch 77595-35. Order control accessories on facing page.
What's Included:
6-ft (1.8-m) line cord with IEC 320/CEE22 socket. These models are shipped with country-specific plug/cord set; please specify ultimate destination when ordering.
Specifications
Product Type
Pump Drive
Max flow rate (mL/min)
1700
Min flow rate (mL/min)
0.18
Control type
Analog Variable speed
Power (Amps)
2.2 at 115 VAC
1.1 at 230 VAC
Motor (hp)
1/10
Motor (watts)
75
Reversible motor
Yes
rpm
3 to 300
IP rating (Drive)
IP33
Dimensions (Drive)
10.8" x 8.3" x 8.5"
(27.4 x 21.1 x 21.6 cm)
Speed control
±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
No. of Pump heads accepted
2
Type of Pump heads accepted
Standard, Easy-Load, Easy-Load II, Easy-Load 3, High Performance, Cartridge, Multichannel, PTFE Tubing, PTFE Diaphragm
Manufacturer number
07528-20
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration for Peristaltic Pump Drive
EW-17050-01
at $115.00 USD/EACH
Usually ships in 8 days.
Add to Cart
Technical Resources
Instruction Manual--English (1.7 MB)
Masterflex
L/S
Variable-Speed Analog Console Drive Brochure
Virtual Catalog (PDFs)
13-14 Catalog
More info
Controladores Masterflex L/S de Precision y Velocidad Variable (folleto en español) (425 KB)
Related technical articles (Roll over for summary)
Masterflex® and the Human Heart: How One Pump is Supporting Research for the Other
A Sweet Success: Famous Maple Syrup Producer Enhances Production with Masterflex® B/T® Variable-Speed Pump
Technical Expertise: How Cole-Parmer Application Specialists Respond to Customers' Needs
Show All...
Reducing Pulsation
Peristaltic Tubing Pumps in Dispensing Applications
Masterflex Tubing: Questions and Answers
Masterflex Tubing Pump Systems FAQs
Tygon® Flexible Plastic Tubing to Tubing Solvent Sealing
Peristaltic Pumps Not Just for Labs Anymore
Tubing Selection Guide For Viscous Fluids
Pumping Abrasives
Pumping Viscous Fluids
Peristaltic Pumps Offer Custom Fluid Solutions
Peristaltic Pumps Offer Economical Multichannel Pumping
Reducing Pulsation in Peristaltic Pumping Systems
Pumping Abrasives
Masterflex Systems: Questions and Answers
High-Performance Masterflex Pump Head
Six Ways to Simplify Fluid Transfer
Masterflex Pump FAQs
Masterflex Tubing Formulations
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Related media
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title