United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 3
of
4 >
Representative photo only
Masterflex L S economy variable speed drive 20 to 600 rpm 115 VAC
Click to enlarge
More Product Details
Specifications
Calibrations
Technical Resources
Instruction manual
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-07554-90
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17050-01
NIST-Traceable Calibration for Peristaltic Pump Drive
$115.00 USD/EACH/EACH
Usually ships in 8 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Masterflex Peristaltic Pumps
>
Masterflex L/S Tubing Pumps
>
Masterflex L/S Drives
>
L/S Analog Variable-Speed Drives
>
L/S Economy Variable-Speed Console Drives
Share
|
Masterflex L/S economy variable-speed drive, 20 to 600 rpm, 115 VAC.
EW-07554-90
$638.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
An economical and versatile variable-speed drive with a wide flow range
Separate single-turn speed control and on/off switch with green power indicatormaintain speed setting while turning pump on/off.
The 1/20-hp unidirectional motor has soft start and back EMF for ±5% speed control.
Stackable painted steel housing is IP22 rated for protection from vertically falling water.
115 VAC models are UL and cUL listed; 230 VAC models are CE marked in compliance with European Community standards.
6-ft (1.8-m) line cord; 115 VAC: U.S. standard plug; 230 VAC: IEC 320/CEE22 socketpumps are shipped with cord/plug set specific to destination country.
Our lowest priced variable-speed drives are a perfect upgrade from a fixed-speed drive. These drives include a wider rpm range, ±5% speed regulation, and UL, cUL, and CE listings. For added versatility, these drives accept the L/S
®
pump head styles specified below.
Specifications
Product Type
Pump Drive
Max flow rate (mL/min)
2900
Min flow rate (mL/min)
1.2
Control type
Variable speed
Flow Chart
Click to View
Power (Amps)
1.5
Motor (hp)
1/20
Motor (watts)
37
rpm
20 to 600
IP rating (Drive)
IP22
Dimensions (Drive)
9"L x 7"W x 6"H
(22.9 x 17.8 x 15.2 cm)
Speed control
±5%
No. of Pump heads accepted
1
Type of Pump heads accepted
Standard, Easy-Load, Easy-Load II, Quick Load
Brand
Masterflex
Model
07554-90
Manufacturer number
07554-90
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration for Peristaltic Pump Drive
EW-17050-01
at $115.00 USD/EACH
Usually ships in 8 days.
Add to Cart
Technical Resources
Instruction Manual (270 KB)
Instruction Manual (215 KB)
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Masterflex® and the Human Heart: How One Pump is Supporting Research for the Other
A Sweet Success: Famous Maple Syrup Producer Enhances Production with Masterflex® B/T® Variable-Speed Pump
Technical Expertise: How Cole-Parmer Application Specialists Respond to Customers' Needs
Show All...
Reducing Pulsation
Peristaltic Tubing Pumps in Dispensing Applications
Masterflex Tubing: Questions and Answers
Masterflex Tubing Pump Systems FAQs
Tygon® Flexible Plastic Tubing to Tubing Solvent Sealing
Peristaltic Pumps Not Just for Labs Anymore
Tubing Selection Guide For Viscous Fluids
Pumping Abrasives
Pumping Viscous Fluids
Peristaltic Pumps Offer Custom Fluid Solutions
Peristaltic Pumps Offer Economical Multichannel Pumping
Reducing Pulsation in Peristaltic Pumping Systems
Pumping Abrasives
Masterflex Systems: Questions and Answers
High-Performance Masterflex Pump Head
Six Ways to Simplify Fluid Transfer
Masterflex Pump FAQs
Masterflex Tubing Formulations
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title