log in or register
Masterflex Linkable Instrument Control Software, CD-ROM
EW-07551-70
$252.00 USD/EACH
  • Control up to 25 Masterflex® L/S® 07551-series drives or Cole-Parmer Servodyne mixers directly from your PC
  • Interface the 07551-series computer-compatible drives via either RS-232 or USB connection
  • Customize the software to meet your application needs: dispensing, mixing/diluting, flow proportioning, and single/multiple slope gradients; program each pump to automate up to 50 steps.
  • Balance interface allows you to dispense off of a balance and also features weight-ratio dispensing; use of multiple balances is enabled with multiple COM ports on your PC
  • Logging options let you log at specific intervals, at the end of each block, or at the end of each run
  • Complies with 21 CFR Part 11: password protection and authorized user list, file encryption permits authorized user access only.
  • Windows®-based software has familiar pull down menus and mouse-selectable icons.
For PC interface via RS-232 port and for daisy-chaining drive to drive use DB9 M/F cable 22050-54; for PC interface via USB port use USB Type A/B cable 22050-60.
Software is supplied in CD-ROM format; compatible with Windows 95/98/NT/2000/XP; Windows XP is recommended for best performance.
Product Type Software
Manufacturer number 07551-70
Brand Masterflex
Accessories
Qty/Price
Availability
at $11.00 USD/EACH
Usually ships in 3 days or sooner.
at $39.00 USD/EACH
In stock.
at $10.00 USD/EACH
In stock.
Technical Resources
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title