United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 4
of
15 >
Representative photo only
Masterflex Standard pump head for L S 14 tubing PC housing CRS rotor
Click to enlarge
More Product Details
See
Upgraded
model
Specifications
Accessories
Replacement parts
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Related media
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-07014-20
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Masterflex Peristaltic Pumps
>
Masterflex Pump Heads
>
Masterflex L/S Pump Heads
>
L/S Standard Pump Heads
>
L/S Standard Pump Heads for Precision Tubing
Share
|
Masterflex Standard pump head for L/S 14 tubing, PC housing/CRS rotor.
EW-07014-20
$135.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Ideal for fluid transfer applications requiring high accuracy and repeatability
Standard heads are precision molded to accommodate a single size of tubinguse for common fluid transfer applications requiring accuracy and excellent repeatability.
Available with clear polycarbonate (PC) housing with either cold-rolled steel (CRS) or stainless steel (SS) rotors; CRS offers economy, while SS offers better chemical resistance and durability
The –52 models with opaque black polyphenylene sulfide (PPS) housing and SS rotors offer the best overall chemical resistance.
Pump head is compact and simple in design with same side tubing entry/exitperfect for space-limited applications.
Standard heads are compact and easy to integrate into your OEM applications. Call for more information on our comprehensive OEM services.
For stacking heads, o
This economical pump head with its classic, patented design is ideal for accurate and repeatable fluid transfer applications. Choose Standard pump head for precision tubing for common fluid transfer applications or Standard pump head for high-performance precision tubing for better pressure generation, suction lift, and the ability to pump viscous fluids.
What's included:
tubing loading key, single-channel mounting hardware, and a 15" (38-cm) length of silicone pump tubing.
Hot Tips:
The design of the standard pump head enables the most precise dispensing accuracy of any MASTERFLEX
®
pump head.
Use only MASTERFLEX
®
tubing with MASTERFLEX pumps to ensure optimum performance. Use of other tubing may void applicable warranties.
Specifications
Product Type
Pump Head
mL/rev
0.21 mL/min
Accepted tubing
L/S® 14
Rotor material
Cold-rolled steel
Housing material
Polycarbonate
Pressure Chart
Click to View
Rollers
Three; cold-rolled steel
Occlusion
Fixed
Dimensions
2-3/4"W x 3"H x 2-1/2"D
(7 x 7.6 x 6.4 cm)
Flow Chart
Click to View
Brand
Masterflex
Manufacturer number
07014-20
Accessories
Qty
/
Price
Availability
Stainless steel mounting hardware; mounts two (2) Masterflex L/S Easy-Load pump heads.
EW-07013-05
at
$34.50 USD/SET of 1
In stock.
Add to Cart
Stainless steel mounting hardware; mounts three (3) Masterflex L/S Easy-Load pump heads.
EW-07013-08
at
$37.00 USD/SET of 1
In stock.
Add to Cart
Stainless steel mounting hardware; mounts four (4) Masterflex L/S Easy-Load pump heads.
EW-07013-09
at
$41.50 USD/SET of 1
In stock.
Add to Cart
Upgraded Model
Qty
/
Price
Availability
Masterflex L/S Easy-Load II pump head for precision tubing, fixed occlusion, CRS rotor.
EW-77200-50
at $228.00 USD/EACH
In stock.
Add to Cart
Replacement parts
Qty
/
Price
Availability
Stainless steel mounting hardware; mounts one (1) Masterflex L/S Easy-Load pump head.
EW-07013-04
at $30.50 USD/SET of 1
In stock.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Masterflex® and the Human Heart: How One Pump is Supporting Research for the Other
Pump Head Torque Requirements
Pump Head Torque Requirements
Show All...
Occlusion: Questions and Answers
L/S® Pump Head Flow Performance
A Sweet Success: Famous Maple Syrup Producer Enhances Production with Masterflex® B/T® Variable-Speed Pump
Technical Expertise: How Cole-Parmer Application Specialists Respond to Customers' Needs
Reducing Pulsation
Peristaltic Tubing Pumps in Dispensing Applications
Masterflex Tubing: Questions and Answers
Masterflex Tubing Pump Systems FAQs
Tygon® Flexible Plastic Tubing to Tubing Solvent Sealing
Peristaltic Pumps Not Just for Labs Anymore
Tubing Selection Guide For Viscous Fluids
Pumping Abrasives
Pumping Viscous Fluids
Peristaltic Pumps Offer Custom Fluid Solutions
Peristaltic Pumps Offer Economical Multichannel Pumping
Reducing Pulsation in Peristaltic Pumping Systems
Pumping Abrasives
Masterflex Systems: Questions and Answers
High-Performance Masterflex Pump Head
Six Ways to Simplify Fluid Transfer
Masterflex Pump FAQs
Masterflex Tubing Formulations
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Related media
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title