United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 6
of
12 >
Representative photo only
Mechanical Convection Oven 5 3cu ft 230VAC
Click to enlarge
More Product Details
Specifications
Technical Resources
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-52402-18
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Heating Equipment
>
Ovens
>
Mechanical Convection Ovens
>
Lab Companion Mechanical Convection Ovens
Share
|
Sorry, this item is now discontinued but has a direct replacement.
You can compare specifications to the direct replacement EW-52402-82
,
browse similar items
or
Contact us for more information.
Mechanical Convection Oven 5.3cu.ft; 230VAC
EW-52402-18
$2,981.00 USD/EACH
Discontinued
Qty
:
Save to shopping list
High air exchange and short rise time for material with a high moisture content
Forced-air convection with a noiseless fan ensures good mixing and strong dispersion
Corrosion-resistant Incoloy
®
sheath heating element and stainless steel interior
Foamed silicone rubber door gasket creates a tight seal for excellent insulation
Mechanical convection ensures fast heat-up and uniform temperature throughout chamber for greater drying efficiency. Ovens are programmable for delayed start and stop. Timer offers you the convenience of unattended operation. The dual-wall door opens 180° for greater access to the chamber. Rounded inner chamber corners offer easy cleaning and superior air flow. Two 1.5” (3.8 cm)-dia adjustable top ventilation slides control inner air and vapor circulation.
What's Included:
two stainless steel wire shelves and power cord.
Specifications
Product Type
Mechanical Convection Oven
Capacity (Liters)
150.043
Capacity (cu ft)
5.3
Chamber size (" W)
22
Resolution
1°C
Chamber size (" H)
23.6
Chamber size (" D)
18
Minimum temperature (º C)
Ambient ±10
Maximum temperature (º C)
250
Minimum temperature (º F)
50
Maximum temperature (º F)
482
Power (VAC)
230
Temp uniformity
±1°C at 100°C
Temp control accuracy
± 1° C
Rise time
100°C within 45 min
Display
Digital LED
Control type
PID microprocessor
Power (Hz)
50/60
No. of shelf positions
16
No. of shelves
2 (included)
CE Compliance
Yes
Brand
Lab Companion
Construction
Stainless steel interior
Overall dimensions
28.7"x38.3"x29.2"
Model
AAH12175K
Manufacturer number
AAH12175K
Technical Resources
Related technical articles (Roll over for summary)
Taking the “Dry” Out of Moisture Determination
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title