Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
 

Medcom Radalert 100 Nuclear Radiation Monitor for measuring alpha, beta, gamma, and X-rays

EW-50511-03
$600.00 USD/EACH

  • Measures alpha, beta, gamma, and x-radiation
  • Digital LCD display
  • User-adjustable alert levels with audio/visual alarm indicators
  • Counts per minute, mR/hr, or accumulated count
  • Optional readout in mSv or microsievert (mSv is millisievert) - mSv is special order (replaces mR/hr)
  • Geiger Mueller detector
  • Interfaces to a computer or data logger
  • Monitors personal radiation exposure
  • Monitors an area or perimeter
  • Detects radiation leaks
  • Checks for contamination
  • Finds radioactive items in the workplace or home
  • Checks for radioactive deposits in the ground
  • Checks for X-ray scatter
  • Locates spill and lost materials
  • Long battery life
  • Ensures regulatory compliance
Many police departments, fire departments, and other emergency response agencies as well as hospitals, laboratories, and industries use the Radalert 100. The digital display shows readings in your choice of counts per minute (up to 100,000 cpm) or mR/hr (up to 100 mR/hr) or in accumulated counts. A red LED blinks and a beeper chirps with each count. An audible alert sounds when the radiation reaches a user-adjustable level.A personal radiation detector is one of the many devices lab workers use to keep them safe. Exposure to radiation through contamination or irradiation can be a significant hazard. Even small amounts over a long time can increase the risk of cancer. In emergency situations, radiation can cause illness, burns, or even death. In nuclear disasters, both the radiation itself and the radioactive ash can be lethal. If a radiation dose is high and penetrates the body, Acute Radiation Syndrome (ARS) may occur. That’s why a personal radiation detector is used to prevent harm. A personal radiation detector monitors the environment to alert users to the presence of radioactive material. Depending on the type (gamma radiation detector, alpha radiation detector, radiation dosimeters), they can detect and measure types of radiation, levels of radiation, counts, accumulated dose, and dose rates. Cole-Parmer offers several low-level sensitive handheld and table-top units with varying features, including documentation capabilities, timers, and ergonomic design. General-purpose detectors are available with uncompensated halogen-quenched sensors for measuring alpha, beta, gamma, and X-ray radiation; select meters have energy-compensated sensors for increased sensitivity to gamma and X-ray radiation. Personal, pen-style dosimeters are also an option. If you have questions about purchasing a personal radiation detector contact a Cole-Parmer Technical Expert today.
What's Included:
Carrying case and one 9V battery
Product Type Radiation detector
Sensitivity 1000 cpm/mR/hr referenced to Cs-137
Sensor Halogen-quenched Geiger-Mueller detector
Mica end window density is 1.5 to 2.0 mg/cm2
Side wall is 0.012" stainless steel
Detects alpha, beta, gamma and X-radiation
Range mR/hr: .001-110; CPM: 0-350,000
µSv/hr: .01-1,100; CPS: 0-3,500;
Total: 0-9,999,000 counts;
Timer: up to 40 hours
Calibration Celsium 137 (gamma)
Display 4-digit liquid crystal display with mode indicators
Alert level User-adjustable alert level to 50 mR/hr and 160,000 cpm
Count light Red LED flashes with each count
Accuracy ±10% typical; ±15% max. (mR/hr & mSv/hr modes)
Dimensions 149 x 81 x 30 mm (5.9" x 3.2" x 1.2")
Output Stereo 3.5 mm jack sends counts to computers, data loggers, other CMOS compatible devices and head-phones
Brand Medcom
Manufacturer number RADALERT 100
Model RADALERT 100
Power One 9-volt alkaline battery; average battery life is 2160 hours at normal background, 625 hours at 1 mR/hr with beeper off

Accessories
Qty/Price
Availability

at $25.60 USD/PACKAGE of 4

In stock.




 
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title