Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

Melting Point Standards

EW-03012-93
$435.00 USD/EACH

Usually ships in 4 days.
Qty

  • High-resolution camera captures the melting point-no operator needs to be present
  • Captures a video image of the melting process with the use of a PC
  • Conforms to US Pharmacopeia (USP) and GLP requirements
This digital unit automatically determines the melting point of chemical substances. A built-in camera continuously captures real-time images of the samples; digital image processing determines when melting occurs. Intuitive setup menu and front-panel touch screen make operation quick and easy; simply select the start temperature, remp reate, stop temperature, and press start. PID-controlled temperature ramping provides an extremely linear increase in temperature. Audible and visual alarms indicate temperature stabilization, end of melt, and temperature outside of low/high limits that are user selected. Analyze up to three samples simultaneously.

View graphical data of the melting event on the front-panel display, or display real-time, high-resolution images of your samples by connecting the unit to a PC through the USB connection. Included MeltView software allows you to play back a melting event as a continuous movie or view it frame-by-frame for detailed analysis. Print out a hard copy of text and graphs using the RS-232 port and the optional printer (sold seperately). Printouts include technician identification and space for a signature.

For samples that melt in a unique way, such as samples that explode, boil away, or bubble up the tube, a wide observation window with a illuminated magnification lens allows manual observation of melting point.

Additional features include three user-configurable timers that can be used when the instrument is not needed for melting point determinations, and an external RTD input (2-, 3-, or 4-wire 100ΩPt) to use the unit as a thermometer when not in use for melting point.





Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title