United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 2
of
3 >
EW-05618-64
Metalex Paddle-Wheel Flow Sensors
Click to enlarge
More Product Details
Specifications
Accessories
Calibrations
Build a complete system
Technical Resources
Instruction manual
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-05618-64
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17080-12
NIST-Traceable Calibration; Liquid Flowmeter, 1 ccm to 300 LPM water
$250.00 USD/EACH/EACH
Usually ships in 19 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Flow, Level & Valves
>
Flowmeters
>
Paddle-Wheel / Impeller Flowmeters
>
+GF+ Signet Flowmeter System
>
Metalex Paddle-Wheel Flow Sensors
Share
|
Metalex Paddle-Wheel Flow Sensors, for use with mini-tap fitting; 2-1/2"
EW-05618-64
$614.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Stainless steel body withstands high pressures and temperatures
Use in agressive liquid flow applications
Use these rugged sensors to monitor the flow of liquid ammonia, turbine steam condensate, and more—wherever strength and chemical resistance are required.
Open-core rotor is cavitation free at flow rates from 1.5 to 20 ft/second. Easy maintenance—sensors can be replaced in minutes.
METALEX flow sensors are compatible with all of our battery- and line-powered flowmeters, accumulators, and controllers. Sensors must be used with the fitting type specified in the table below—installation fittings sold separately.
METALEX sensors feature CD4MCU rotor; Fluoroloy B rotor bearings; and 316 stainless steel body, rotor shaft, and shaft retainers.
What's Included:
25-foot signal cable
Specifications
Product Type
Paddle Wheel Flowmeter
Description
Paddle Wheel Flowmeter
Max operating temperature (º F)
300
Max operating temperature (º C)
149
Max pressure
1500 psi
Repeatability
±0.5% full-scale
Brand
+GF+ Signet
Model
P525-2
Manufacturer number
P525-2
Build a complete system
1
.
+GF+ Signet Rotor-X Paddle-Wheel Flow Sensors
Details
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; Liquid Flowmeter, 1 ccm to 300 LPM water
EW-17080-12
at $250.00 USD/EACH
Usually ships in 19 days.
Add to Cart
Technical Resources
Instruction Manual (81.7 KB)
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Paddlewheel Flowmeters: High flow system accuracy on a modest budget
Installing Your Paddle-Wheel Flow Sensor
Velocity-Profile Deviations Influence Flowmeter Performance
Show All...
Selecting the Right FlowmeterPart 1
Selecting the Right FlowmeterPart 2
Electron zap turns flimsy plastic into sturdy shrink wrap
Basics of Cartridge Filtration
Bitter Brew
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title