log in or register
Metallic Tubing Cutter, 110-120/220-240 V
EW-95649-00
$960.00 USD/EACH
  • Clean, burr-free cuts improve chromatographic reproducibility
  • Cuts metallic tubing faster with more precision
  • Assures zero dead volume tubing connections in chromatography applications
  • Eliminates sample dispersion caused by improper deburring of tubing
Eliminate burrs and uneven cuts on stainless steel and copper tubing for chromatography applications. Clamp vice on swing arm holds tubing securely in place while the abrasive cutting wheel produces a square cut on tubing. Protective shield covers the abrasive wheel for safe operation. Use with any HPLC system or for chromatography pilot scale plumbing.
Product Type Tubing Cutters
Accessories
Qty/Price
Availability
at $30.00 USD/EACH
Usually ships in 11 days.
at $46.00 USD/PACKAGE
In stock.
Technical Resources
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title