Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

Methanol, Anhydrous Acs 4L acs poly gls

EW-89100-08
$68.00 USD/EACH

quantity discounts Quantity Discounts
Order Discount Price
[X]
4 10% $61.20/EACH
8 15% $57.80/EACH
16 20% $54.40/EACH
Usually ships in 5 days.
Qty

This item is considered a hazardous material.Please note: there may be additional shipping restrictions like freight collect or no priority air service, extra freight charges may apply. This item(s) may be restricted from shipping to a residential address, sold to private individuals or exported from the United States. For additional information or questions please call us at 1-800-323-4340 (US) or 847-549-7600 (International).
Product Type Chemical Reagents
Purity grade ACS
CAS Number 67-56-1
Form Liquid
Formula weight 32.04
Empirical formula CH4O
Linear formula CH3OH
Synonyms Methyl Alcohol
Packaging Poly Bottle
Storage symbol Red
UN number UN 1230
Hazard symbol Flammable
Quantity (mL) 4000
DOT information DOT 3 PG II
DOT description Methanol
Brand Cole-Parmer
Manufacturer number 910008
Quantity 4 L





Technical Resources
Certificate of Analysis
Enter lot number

or click to view a Sample C of A
 
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title