dummy
EW-94190-05
Call 1-888-358-4705 for availability.
Qty:
- Easy-to-use, compact endpoint titrator
- Select a method from the menu based on your application
- Press one button to begin operation
Titrator is perfect for acidity and alkalinity determination. All parameters are already optimized so no further method development is needed. Press the Run button to start the method—titrator remembers the last method you ran. An online graph safely guides you to the result. Select normal speed for typical samples, fast speed when you need quick results, or cautious speed for critical samples.
Mettler’s well-proven sample beaker attachment is easy to use—even the very first time. Just clip in and you are ready to go. Additional performance features include password protection for custom methods, soft-key memory for up to three of your most used methods, pictogram keys for commonly used functions such as stirrer activation, burette rinsing, and manual pH mode. Titrator has a 100-sample memory with statistics and can produce a GLP-conforming printout.
What's Included:
magnetic stirrer with stirring bars, pH electrode and cable, 20-mL burette, titration stand with two titration beakers, and printer cables.
| Product Type |
Acid-Base/Alkalinity Titrators |
| CE Compliance |
Yes |
| Brand |
Mettler Toledo |
| Manufacturer number |
DL15 |
| Model |
DL15 |
at $49.00 USD/PACKAGE
Usually ships in 3 days.
at $25.00 USD/EACH
Usually ships in 14 days.
at $1,041.00 USD/EACH
Usually ships in 10 days.
at $300.00 USD/EACH
In stock.
Technical Resources
Related technical articles (Roll over for summary)
Show Less...
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.