United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 2
of
5 >
EW-11334-48
METTLER TOLEDO Moisture Determination Balance, 54 g x 0.01%
Click to enlarge
More Product Details
Specifications
Accessories
Related products
Calibrations
Technical Resources
Virtual Catalog (PDFs)
More info
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-11334-48
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17101-52
NIST Traceable Report With Data, Balances/Scales - all types
$140.00 USD/EACH/EACH
Usually ships in 7 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Balances & Scales
>
Moisture Balances
>
Mettler Toledo Moisture Balances
Share
|
Mettler Toledo Moisture Determination Balance, 54 g x 0.01%
EW-11334-48
$4,347.00 USD/EACH
Usually ships in 7 days.
Qty
:
Add to Cart
Save to shopping list
Designed for simple laboratory applications and moisture determinations
Suitable for ISO, GLP, and GMP protocols
Adjustable two-point calibration of dryer and balance
Drying time of 1 to 480 minutes
Ergonomic keyboard features large keys and backlit LCD
Use factory settings or preselect test parameters for optimal results
Analyzers reach temperature quickly with exceptional heating uniformity
When testing is complete, unit cools down quickly for greater sample throughput and user safety
Three selectable switch-off criteria allow you to end a drying program based on a weight loss preset amount of time or manually
Battery backup saves data in the event of a power failure
Built-in overheating protection protects analyzer
RS-232 interface connects analyzer to an external printer or PC
Rugged design, basic functionality, and simple operation for daily routine inspection
No programming needed the end-point of the analysis is determined automatically
Easy to use just open the cover and the moisture analyzer leads you step by step through the measurement
Economically priced and designed for simple laboratory applications and moisture determinations in production areas. Ergonomic keyboard features large keys and backlit LCD. Use factory settings or pre-select test parameters for optimal results. Analyzers reach temperatures quickly with exceptional heating uniformity, ensuring excellent reproducibility. When testing is complete, unit cools down quickly for greater sample throughput and user safety. Three selectable switch-off criteria allow you to end a drying program based on a weight loss per unit of time, after a preset amount of time has elapsed, or manually. Battery backup saves data in the event of a power failure. Built-in overheating potection protects analyzer. RS-232 interface connects analyzer to an external printer or PC.
Available in the U.S. and Puerto Rico
What's Included:
80 aluminum sample pans, sample holder, sample handler, specimen sample (absorbent cellulose disc), draft shield, and power cable
Specifications
Product Type
Moisture Balances
Calibration
External
Capacity (g)
54
Capacity (oz)
1.9
Heater type
Halogen
Housing dimensions
9" x 6" x 14-1/4" (22.9 x 15.2 x 36.2 cm)
Pan size (dia)
3-5/8" (9.2 cm)
Power (VAC)
115
Readability
1 mg
Repeatability
±0.10% (2 g sample)
Temperature range (º C) (low)
50
Temperature range (º C) (high)
200
Timer (min)
1 to 480
Weighing units
% moisture, % solid, % moisture dry, mg weight loss, or grams of residue
Output
RS-232
Brand
Mettler Toledo
Accessories
Qty
/
Price
Availability
12 Months Extended Warranty for All Mettler Toledo Balances
EW-11332-12
at
$218.00 USD/EACH
Usually ships in 5 days.
Add to Cart
24 Months Extended Warranty for All Mettler Toledo Balances
EW-11332-24
at
$343.00 USD/EACH
Usually ships in 13 days.
Add to Cart
Related products
Qty
/
Price
Availability
Cole-Parmer SpatulaBalance
, 300 g x 0.1 g
EW-01019-11
at $43.00 USD/EACH
In stock.
Add to Cart
Disposable foil weighing pans, pack of 50.
EW-11334-93
at $85.00 USD/PACKAGE
In stock.
Add to Cart
Calibrations
Qty
/
Price
Availability
NIST Traceable Report With Data, Balances/Scales - all types
EW-17101-52
at $140.00 USD/EACH
Usually ships in 7 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
More info
Mettler Toledo Moisture Analyzer Brochure (306 KB)
Related technical articles (Roll over for summary)
Taking the “Dry” Out of Moisture Determination
Gravitational Effects on Balance
Weighing Unit Conversion Table
Show All...
Common Balance Terms
Equipping a Laboratory: A Study in Customer Service Excellence
Temperature FAQs
Temperature Instrument Ranges
Getting a Handle on Temperature
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title