United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 3
of
3
EW-11334-79
METTLER TOLEDO NewClassic ML Analytical Balance, 220g X 0.1mg. Representative Photo Only
Click to enlarge
More Product Details
Specifications
Accessories
Related products
Calibrations
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-11334-79
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17101-52
NIST Traceable Report With Data, Balances/Scales - all types
$140.00 USD/EACH/EACH
Usually ships in 7 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Balances & Scales
>
Analytical Balances
>
Mettler Toledo Analytical Balances
>
Mettler Toledo NewClassic ML Analytical Balances
Share
|
Mettler Toledo NewClassic ML Analytical Balance, 220g X 0.1mg
EW-11334-79
$3,435.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Designed for mobility and easy operation
Advanced MonoBloc™ weighing technology provides fast and accurate results
Direct connection to a PC via RS232 slot allows convenient transfer of weighing results to Excel®
Built-in icon-based application programs including piece counting, percent weighing, dynamic weighing, statistics, statistics, formulation, totaling, free factor, and recall
AC or battery operable for up to 8 hours with 8 AAA batteriesideal for work in field or whenever a power supply is hard to find
Overload Protection guards the weighing cell against excess weight overload
SmartTrac™ software displays how much of the entire weighing range has been used
NewClassic ML balances have everything a basic balance needs: an ergonomic design with a large, backlit display, easy-to-clean glass draft shield, an intuitive user interface, multiple built-in applications, battery operation for full mobility, and excellent weighing performance.
What's Included:
120 VAC power adapter
Specifications
Product Type
Analytical Balances
Calibration
Internal
Capacity (g)
220
Capacity (oz)
8
Housing dimensions
19.1 x 33 x 29 cm
Dimensions (" W)
7-1/2
Dimensions (" H)
13
Dimensions (" D)
11-1/2
Display
Backlit Display
Pan size (dia)
3.5" (9 cm)
Readability (mg)
0.1
Linearity (mg)
±0.2
Repeatability (mg)
±0.1
Weighing units
g, kg, mg, ct, lb, oz, ozt, GN, dwt, momme, mesghal, tael, tical, tola, baht
Stabilization time
2 sec
Stability filters
three filters to match weighing environment
Power (VAC)
120
Battery
8 AAA batteries (not included)
Languages
English, German, French, Spanish, Italian, Polich, Czech, Hugarian
Output
RS-232
Brand
Mettler Toledo
Accessories
Qty
/
Price
Availability
Batteries, 1.5 V, AAA, 12/pack
EW-09376-00
at
$21.25 USD/PACKAGE of 12
In stock.
Add to Cart
Related products
Qty
/
Price
Availability
Cole-Parmer SpatulaBalance
, 300 g x 0.1 g
EW-01019-11
at $43.00 USD/EACH
In stock.
Add to Cart
Calibrations
Qty
/
Price
Availability
NIST Traceable Report With Data, Balances/Scales - all types
EW-17101-52
at $140.00 USD/EACH
Usually ships in 7 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Optimizing Weighing Performance
Gravitational Effects on Balance
Weighing Unit Conversion Table
Show All...
Common Balance Terms
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title