United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 2
of
3 >
EW-11334-55
METTLER TOLEDO NewClassic MS Analytical Balance, 220g X 0.1mg. Representative Photo Only
Click to enlarge
More Product Details
Specifications
Related products
Calibrations
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-11334-55
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17101-52
NIST Traceable Report With Data, Balances/Scales - all types
$140.00 USD/EACH/EACH
Usually ships in 7 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Balances & Scales
>
Analytical Balances
>
Mettler Toledo Analytical Balances
>
Mettler Toledo NewClassic MS Analytical Balances
Share
|
Mettler Toledo NewClassic MS Analytical Balance, 220g X 0.1mg
EW-11334-55
$4,523.00 USD/EACH
Usually ships in 7 days.
Qty
:
Add to Cart
Save to shopping list
Fast settling time and consistent results for routine to complicated weighing procedures
Fully automatic time- and temperature-controlled internal calibration adjustment
Advanced MonoBloc™ weighing technology provides fast and accurate results
Durable high-grade chemical-resistant die-cast aluminum housing
Overload Protection guards the weighing cell against excess weight overload
SmartTrac™ software displays how much of the entire weighing range has been used
Direct connection to a PC or printer via RS232 or USB slot allows convenient transfer of weighing results
High-contrast display (HCD) and intuitive menu for simple operation
Programmable Smart Keys for shortcut access to preferred applications
These balances feature a robust housing made of high quality chemical-resistant aluminum that protects the sensitive sensor from environmental influences and impacts. The proven MonoBloc weighing technology and FACT – fully automatic time- and temperature-controlled internal adjustment – provide accurate, repeatable results for everyday weighing tasks. A new High Contrast Display with big numbers, clear symbols and an intuitive menu with new programmable SmartKeys allows users to operate the balance quickly and easily with one touch operation.
What's Included:
Draft shield, protective in-use cover, and AC power adapter
Specifications
Product Type
Analytical Balances
Calibration
Internal
Capacity (g)
220
Capacity (oz)
8
Housing dimensions
20.4 x 34.8 x 34.7 cm
Dimensions (" W)
8
Dimensions (" H)
13-3/4
Dimensions (" D)
13-5/8
Display
High Contrast Display (HCD)
Pan size (dia)
3-1/2" (9 cm)
Readability (mg)
0.1
Linearity (mg)
±0.2
Repeatability (mg)
±0.1
Weighing units
g, kg, mg, ct, lb, oz, ozt, GN, dwt, mom, msg, tlh, tls, tlt, tola, baht
Stabilization time
2 sec
Stability filters
three filters to match weighing environment
Power (VAC)
120
Battery
None
Languages
English, German, French, Spanish, Italian, Polich, Czech, Hugarian
Output
RS-232 / USB
Brand
Mettler Toledo
Related products
Qty
/
Price
Availability
Cole-Parmer SpatulaBalance
, 300 g x 0.1 g
EW-01019-11
at $43.00 USD/EACH
In stock.
Add to Cart
Calibrations
Qty
/
Price
Availability
NIST Traceable Report With Data, Balances/Scales - all types
EW-17101-52
at $140.00 USD/EACH
Usually ships in 7 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Optimizing Weighing Performance
Gravitational Effects on Balance
Weighing Unit Conversion Table
Show All...
Common Balance Terms
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title